Search
Search

TIMM44| Gene Symbol: TIMM44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015122713.2 | XP_014978199.1 | 2464 | 70 | ||
| GenBank mRNA | JU326879.1 | - | 838 | 70 | ||
| JV568954.1 | - | 833 | 70 | |||
| JV640236.1 | - | 847 | 70 | |||
Set Membership: |
|