Search
Search

CYP3A4| Gene Symbol: CYP3A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001040414.1 | NP_001035504.1 | 164 | 96 | ||
| GenBank mRNA | AY334551.1 | - | 164 | 96 | ||
| AY582531.1 | - | 164 | 96 | |||
Set Membership: |
|