Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ETNPPL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087348.4 | XP_001087348.3 | 768 | 133 | ||
GenBank mRNA | JU323161.1 | - | 559 | 133 | ||
JV452217.1 | - | 558 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |