Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: LAMTOR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257535.1 | NP_001244464.1 | 264 | 117 | ||
GenBank mRNA | JU320556.1 | - | 264 | 117 | ||
JU471004.1 | - | 228 | 117 | |||
JV044392.1 | - | 244 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |