Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2F2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102839.4 | XP_001102839.1 | 677 | 73 | ||
GenBank mRNA | JU470817.1 | - | 682 | 73 | ||
JU474573.1 | - | 682 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |