Search
Search

LRMDA| Gene Symbol: LRMDA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028826439.1 | XP_028682272.1 | 669 | 122 | ||
| XM_028826440.1 | XP_028682273.1 | 673 | 122 | |||
| XM_028826441.1 | XP_028682274.1 | 672 | 122 | |||
| XM_028826442.1 | XP_028682275.1 | 1460 | 122 | |||
| XM_028826443.1 | XP_028682276.1 | 513 | 122 | |||
| GenBank mRNA | JV724786.1 | - | 189 | 122 | ||
Set Membership: |
|