Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084619.4 | XP_001084619.2 | 1891 | 144 | ||
XM_028829264.1 | XP_028685097.1 | 2062 | 144 | |||
XM_028829265.1 | XP_028685098.1 | 2108 | 144 | |||
GenBank mRNA | JU336522.1 | - | 1547 | 144 | ||
JU336523.1 | - | 1547 | 144 | |||
JU477331.1 | - | 1558 | 144 | |||
JU477332.1 | - | 1558 | 144 | |||
JV048325.1 | - | 1542 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |