Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261606.1 | NP_001248535.1 | 739 | 139 | ||
XM_015128312.2 | XP_014983798.1 | 1934 | 139 | |||
GenBank mRNA | JV048326.1 | - | 739 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |