Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194589.2 | NP_001181518.1 | 1056 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |