Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Rh02886963_m1
See other ADGRF3 GE Assays ›
Gene Symbol:
ADGRF3
Gene Name:
adhesion G protein-coupled receptor F3
Gene Aliases:
GPR113
Chromosome Location:
Chr.13: 82631247 - 82672780 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID
Rh02886963_m1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
435.00
Your Price
Online offer:
408.65
408.65
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: ADGRF3
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
XM_015111872.2
XP_014967358.2
1990
104
XM_015111873.2
XP_014967359.2
1967
104
XM_028831872.1
XP_028687705.1
2028
104
XM_028831873.1
XP_028687706.1
1862
104
XM_028831874.1
XP_028687707.1
2007
104
XM_028831875.1
XP_028687708.1
1871
104
XM_028831876.1
XP_028687709.1
1736
104
XM_028831877.1
XP_028687710.1
1636
104
XM_028831878.1
XP_028687711.1
1989
104
XM_028831879.1
XP_028687712.1
1990
104
XM_028831880.1
XP_028687713.1
1961
104
XM_028831881.1
XP_028687714.1
1891
104
XM_028831882.1
XP_028687715.1
1743
104
XM_028831883.1
XP_028687716.1
1510
104
XM_028831884.1
XP_028687717.1
1601
104
XM_028831885.1
XP_028687718.1
1553
104
XM_028831886.1
XP_028687719.1
1587
104
XM_028831887.1
XP_028687720.1
1537
104
XM_028831888.1
XP_028687721.1
1003
104
XM_028831889.1
XP_028687722.1
908
104
XM_028831890.1
XP_028687723.1
1594
104
XM_028831891.1
XP_028687724.1
809
104
XM_028831892.1
XP_028687725.1
1463
104
GenBank mRNA
JU339690.1
-
1344
104
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
G-protein coupled receptor
3' Most
Recommended Assay
Macaca mulatta
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline