Search
Search

USP22| Gene Symbol: USP22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015118820.2 | XP_014974306.1 | 1167 | 83 | ||
| XM_015118821.2 | XP_014974307.1 | 798 | 83 | |||
| XM_028836125.1 | XP_028691958.1 | 1212 | 83 | |||
| GenBank mRNA | JU320229.1 | - | 1041 | 83 | ||
| JU320230.1 | - | 1041 | 83 | |||
| JU474900.1 | - | 978 | 83 | |||
| JU474901.1 | - | 978 | 83 | |||
| JV043391.1 | - | 1147 | 83 | |||
| JV043392.1 | - | 1147 | 83 | |||
Set Membership: |
|