Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00431780_m1
          See other ALX4 GE Assays ›
          Gene Symbol:
          Alx4
          Gene Name:
          ALX homeobox 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 82548959 - 82585531 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00431780_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Alx4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001106553.1 NP_001100023.1 1 - 2 600 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001206.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          pattern specification process
          muscle organ development
          biological_process
          post-embryonic development
          anterior/posterior pattern specification
          limb morphogenesis
          embryonic forelimb morphogenesis
          embryonic hindlimb morphogenesis
          embryonic digit morphogenesis
          regulation of apoptotic process
          positive regulation of transcription from RNA polymerase II promoter
          digestive tract development
          embryonic skeletal system morphogenesis
          skeletal system morphogenesis
          palate development
          RNA polymerase II regulatory region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding
          molecular_function
          transcription factor activity, sequence-specific DNA binding
          sequence-specific DNA binding
          protein heterodimerization activity
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P116819 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 713256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324961 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 602141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861303 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874241 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 177828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 860576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102247 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734437 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 457510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278838 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340365 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 353029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 352010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 107984 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 17461 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 79972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 108319 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116819 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P116819 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P116819 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P253137 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10838071 - -
          Affymetrix Rat Gene 1.0 ST Array 10838072 - -
          Affymetrix Rat Gene 1.0 ST Array 10838074 - -
          Affymetrix Rat Gene 1.0 ST Array 10838070 - -
          Affymetrix Rat Gene 1.0 ST Array 10838073 - -
          Affymetrix Rat Gene 1.1 ST Array 10838069 - -
          Affymetrix Rat Clariom S Assay PSR0300008459.rn.1 TC0300001206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300008460.rn.1 TC0300001206.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300008458.rn.1 TC0300001206.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10838069 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21449314 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1366417 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline