Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00433209_m1
          See other PTK2 GE Assays ›
          Gene Symbol:
          Ptk2
          Gene Name:
          protein tyrosine kinase 2
          Gene Aliases:
          FAK, FRNK, p125FAK
          Chromosome Location:
          Chr.7: 114436419 - 114611334 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00433209_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ptk2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013081.2 NP_037213.1 20 - 21 2254 68
          XM_008765546.1 XP_008763768.1 24 - 25 2422 68
          GenBank mRNA AF020777.1 - 20 - 21 2253 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700003313.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase Recommended Assay Rattus norvegicus Rat Tumor Metastasis Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ptk2

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          microtubule cytoskeleton organization
          angiogenesis
          blood vessel development
          vasculogenesis
          neuron migration
          positive regulation of protein phosphorylation
          cytoskeleton organization
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          JNK cascade
          positive regulation of cell proliferation
          regulation of cell shape
          response to mechanical stimulus
          response to glucose
          response to organic substance
          response to organonitrogen compound
          positive regulation of phosphatidylinositol 3-kinase signaling
          response to organic cyclic compound
          positive regulation of smooth muscle cell migration
          peptidyl-tyrosine phosphorylation
          central nervous system neuron axonogenesis
          negative regulation of cell-cell adhesion
          extracellular matrix organization
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of cell migration
          cellular chloride ion homeostasis
          response to estradiol
          regulation of cell adhesion mediated by integrin
          peptidyl-tyrosine autophosphorylation
          establishment of nucleus localization
          regulation of cell proliferation
          response to drug
          negative regulation of apoptotic process
          endothelial cell migration
          innate immune response
          fat cell differentiation
          regulation of osteoblast differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          positive regulation of vasodilation
          negative regulation of organ growth
          response to arsenic-containing substance
          protein autophosphorylation
          ephrin receptor signaling pathway
          positive regulation of smooth muscle cell proliferation
          positive regulation of phagocytosis
          negative regulation of axonogenesis
          positive regulation of synaptic transmission
          positive regulation of protein kinase B signaling
          negative regulation of synapse assembly
          positive regulation of glial cell proliferation
          growth hormone receptor signaling pathway
          cellular response to transforming growth factor beta stimulus
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of anoikis
          actin binding
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          integrin binding
          protein binding
          ATP binding
          JUN kinase binding
          protein kinase binding
          protein complex binding
          SH2 domain binding
          identical protein binding
          phosphatidylinositol 3-kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1045425 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 963668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912191 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99093 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444486 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756815 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1512 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 219143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392723 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 633436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 377167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 494590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 950096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 112317 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 914363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 484261 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 551530 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163867 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957518 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 253722 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 447593 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 451803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594885 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725767 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776255 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 711449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43121 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 190392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 194343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926198 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 13870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790692 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88206 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445291 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496760 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 717510 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435680 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951590 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234022 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299583 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 307522 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383988 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266937 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 500274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 1016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 251458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 542600 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 657413 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 43400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946638 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163456 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 17581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 325246 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279274 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 529508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912851 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888470 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 676964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249333 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 139650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 682298 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92499 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772576 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487097 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380751 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 879938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 894458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 51010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 753027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 250776 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263484 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 341651 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19425 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 93751 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148428 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159477 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164278 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 146227 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65289 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101154 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145011 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72839 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 101149 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 72145 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145125 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 65355 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 122980 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 67323 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 140429 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 3439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045425 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1045425 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1045425 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P510809 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P836576 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1045425 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P667865 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 104488 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 86241 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 6622 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9482 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40801 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56058 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99056 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9181 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4189 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 105014 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 59227 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24277 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 2784 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77162 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10904391 - -
          Affymetrix Rat Gene 1.0 ST Array 10904405 - -
          Affymetrix Rat Gene 1.0 ST Array 10904396 - -
          Affymetrix Rat Gene 1.0 ST Array 10904413 - -
          Affymetrix Rat Gene 1.0 ST Array 10904410 - -
          Affymetrix Rat Gene 1.0 ST Array 10904409 - -
          Affymetrix Rat Gene 1.0 ST Array 10904392 - -
          Affymetrix Rat Gene 1.0 ST Array 10904404 - -
          Affymetrix Rat Gene 1.0 ST Array 10904383 - -
          Affymetrix Rat Gene 1.0 ST Array 10904393 - -
          Affymetrix Rat Gene 1.0 ST Array 10904412 - -
          Affymetrix Rat Gene 1.0 ST Array 10904411 - -
          Affymetrix Rat Gene 1.0 ST Array 10904403 - -
          Affymetrix Rat Gene 1.0 ST Array 10904402 - -
          Affymetrix Rat Gene 1.0 ST Array 10904385 - -
          Affymetrix Rat Gene 1.0 ST Array 10904381 - -
          Affymetrix Rat Gene 1.0 ST Array 10904380 - -
          Affymetrix Rat Gene 1.0 ST Array 10904388 - -
          Affymetrix Rat Gene 1.0 ST Array 10904389 - -
          Affymetrix Rat Gene 1.0 ST Array 10904397 - -
          Affymetrix Rat Gene 1.0 ST Array 10904390 - -
          Affymetrix Rat Gene 1.0 ST Array 10904387 - -
          Affymetrix Rat Gene 1.0 ST Array 10904408 - -
          Affymetrix Rat Gene 1.0 ST Array 10904407 - -
          Affymetrix Rat Gene 1.0 ST Array 10904400 - -
          Affymetrix Rat Gene 1.0 ST Array 10904382 - -
          Affymetrix Rat Gene 1.0 ST Array 10904386 - -
          Affymetrix Rat Gene 1.0 ST Array 10904401 - -
          Affymetrix Rat Gene 1.0 ST Array 10904384 - -
          Affymetrix Rat Gene 1.0 ST Array 10904395 - -
          Affymetrix Rat Gene 1.1 ST Array 10904377 - -
          Rat Genome 230 2.0 Array 1394338_x_at - -
          Rat Genome 230 2.0 Array 1394338_PM_x_at - -
          Rat Genome 230 2.0 Array 1387875_at - -
          Rat Genome 230 2.0 Array 1387875_PM_at - -
          Affymetrix HT Rat Focus Array 1387875_at - -
          Affymetrix HT Rat Focus Array 1394338_x_at - -
          Affymetrix HT Rat Focus Array 1387875_PM_at - -
          Affymetrix HT Rat Focus Array 1394338_PM_x_at - -
          Affymetrix Rat Clariom S Assay PSR0700025302.rn.1 TC0700003313.rn.1 RTA TC
          Rat Expression Set 230 1387875_PM_at - -
          Rat Expression Set 230 1394338_x_at - -
          Rat Expression Set 230 1394338_PM_x_at - -
          Rat Expression Set 230 1387875_at - -
          Affymetrix Rat Gene 1.0 ST Array 10904377 - -
          Rat Genome U34 Set S83358_s_at - -
          Rat Genome U34 Set rc_AI145420_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21475654 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1359854 - -
          RatRef-12 Expression BeadChip ILMN_68931 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline