Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00435938_m1
          See other PRKG2 GE Assays ›
          Gene Symbol:
          Prkg2
          Gene Name:
          protein kinase, cGMP-dependent, type II
          Gene Aliases:
          GDPKII, cGKII
          Chromosome Location:
          Chr.14: 12217121 - 12313616 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00435938_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkg2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_013012.1 NP_037144.1 15 - 16 1985 99
          GenBank mRNA Z36276.1 - 15 - 16 1985 99

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1400000196.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkg2

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          circadian rhythm
          circadian regulation of gene expression
          peptidyl-serine autophosphorylation
          protein targeting to plasma membrane
          cGMP-dependent protein kinase activity
          ATP binding
          cGMP binding
          protein homodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P123381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953026 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417151 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 293050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 154686 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 93780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 600101 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 929977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 865302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 146201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 25919 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 642388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 501950 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 435453 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 174349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573381 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 569907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946762 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 418849 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669476 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 959705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817183 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189442 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 456061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 739113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422229 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 803502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 150376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807689 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 548409 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 506996 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 78902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64318 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877621 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416787 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 284933 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 925585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647392 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 496265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281366 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106114 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 172556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98378 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98078 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132838 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 109134 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 105151 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99610 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120171 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94629 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33788 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 120985 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168828 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 98256 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P123381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P123381 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P123381 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P123381 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 1563 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55840 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56809 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99673 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 97353 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 30011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90379 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 56065 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10771515 - -
          Affymetrix Rat Gene 1.0 ST Array 10771520 - -
          Affymetrix Rat Gene 1.0 ST Array 10771519 - -
          Affymetrix Rat Gene 1.0 ST Array 10771512 - -
          Affymetrix Rat Gene 1.0 ST Array 10771509 - -
          Affymetrix Rat Gene 1.0 ST Array 10771522 - -
          Affymetrix Rat Gene 1.0 ST Array 10771517 - -
          Affymetrix Rat Gene 1.0 ST Array 10771513 - -
          Affymetrix Rat Gene 1.0 ST Array 10771518 - -
          Affymetrix Rat Gene 1.0 ST Array 10771521 - -
          Affymetrix Rat Gene 1.0 ST Array 10771516 - -
          Affymetrix Rat Gene 1.0 ST Array 10771507 - -
          Affymetrix Rat Gene 1.0 ST Array 10771523 - -
          Affymetrix Rat Gene 1.0 ST Array 10771511 - -
          Affymetrix Rat Gene 1.0 ST Array 10771514 - -
          Affymetrix Rat Gene 1.0 ST Array 10771508 - -
          Affymetrix Rat Gene 1.0 ST Array 10771510 - -
          Affymetrix Rat Gene 1.0 ST Array 10771524 - -
          Affymetrix Rat Gene 1.1 ST Array 10771506 - -
          Rat Genome 230 2.0 Array 1369090_at - -
          Rat Genome 230 2.0 Array 1369090_PM_at - -
          Affymetrix HT Rat Focus Array 1369090_PM_at - -
          Affymetrix HT Rat Focus Array 1369090_at - -
          Affymetrix Rat Clariom S Assay PSR1400001625.rn.1 TC1400000196.rn.1 RTA TC
          Rat Expression Set 230 1369090_PM_at - -
          Rat Expression Set 230 1369090_at - -
          Affymetrix Rat Gene 1.0 ST Array 10771506 - -
          Rat Genome U34 Set Z36276_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21432274 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361898 - -
          RatRef-12 Expression BeadChip ILMN_68009 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline