Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00436469_m1
          See other SEMA3A GE Assays ›
          Gene Symbol:
          Sema3a
          Gene Name:
          sema domain, immunoglobulin domain (Ig), short basic domain, secreted, (semaphorin) 3A
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 18170375 - 18545190 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00436469_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Sema3a
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_017310.1 NP_059006.1 14 - 15 1653 75
          XM_006235989.2 XP_006236051.1 16 - 17 2040 75
          XM_006235990.2 XP_006236052.1 15 - 16 1893 75
          XM_008762702.1 XP_008760924.1 16 - 17 1902 75
          GenBank mRNA X95286.1 - 14 - 15 1653 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002298.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Sema3a

          Panther Classification:

          Molecular Function -

          membrane-bound signaling molecule intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          neuron migration
          regulation of heart rate
          apoptotic process
          axon guidance
          axonal fasciculation
          negative regulation of epithelial cell migration
          facial nerve structural organization
          trigeminal nerve structural organization
          nerve development
          olfactory bulb development
          branchiomotor neuron axon guidance
          gonadotrophin-releasing hormone neuronal migration to the hypothalamus
          negative regulation of axon extension
          ventral trunk neural crest cell migration
          dendrite morphogenesis
          regulation of axon extension involved in axon guidance
          negative regulation of axon extension involved in axon guidance
          axon extension involved in axon guidance
          negative chemotaxis
          axonogenesis involved in innervation
          dichotomous subdivision of terminal units involved in salivary gland branching
          sympathetic ganglion development
          trigeminal ganglion development
          semaphorin-plexin signaling pathway
          sympathetic neuron projection extension
          sympathetic neuron projection guidance
          neural crest cell migration involved in autonomic nervous system development
          semaphorin-plexin signaling pathway involved in neuron projection guidance
          semaphorin-plexin signaling pathway involved in axon guidance
          neural crest cell migration involved in sympathetic nervous system development
          facioacoustic ganglion development
          positive regulation of male gonad development
          positive regulation of neuron migration
          receptor activity
          semaphorin receptor binding
          neuropilin binding
          chemorepellent activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888056 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 848078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 431073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 895429 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 161157 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 794270 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 92637 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 131400 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 866201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 169820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 750563 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567717 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 749917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 66232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 18324 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 746940 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 433868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 120276 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 945740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 716410 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 835711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 58954 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 807624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 47706 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 817613 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 385963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273668 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725223 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 613481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 295962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 909814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439488 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 843798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 70648 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 576475 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144960 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 39096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 589066 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 241352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373135 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 962384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 846312 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233311 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 173289 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109290 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 645677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22366 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658831 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 74320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 709774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 831972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 305330 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 854430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466419 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696553 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330473 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 631081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 850942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498596 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 162907 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 276915 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 495345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 242764 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 430109 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814012 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 508585 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 707114 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60258 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 699474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 811417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 519759 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 911631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 278415 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 126090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795019 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738347 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38715 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 267999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 789211 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274131 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373387 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 109074 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 299053 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 141038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 579029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 202050 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 573201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 94828 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 22529 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 872309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 55605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 630808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73264 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344028 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358202 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 10666 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402017 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 265145 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 720399 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 583231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 917030 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115712 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 169991 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115733 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57686 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 145372 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111476 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119156 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 88566 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55032 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56100 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 130315 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6898 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 141393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 150550 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 21823 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 176766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44637 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137071 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 77113 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96564 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 61762 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 57880 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11908 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11908 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11908 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11908 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60315 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 85261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62151 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62328 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 96230 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61750 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13498 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 70534 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13474 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15701 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23581 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14437 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 28168 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10860490 - -
          Affymetrix Rat Gene 1.0 ST Array 10860495 - -
          Affymetrix Rat Gene 1.0 ST Array 10860497 - -
          Affymetrix Rat Gene 1.0 ST Array 10860489 - -
          Affymetrix Rat Gene 1.0 ST Array 10860485 - -
          Affymetrix Rat Gene 1.0 ST Array 10860486 - -
          Affymetrix Rat Gene 1.0 ST Array 10860484 - -
          Affymetrix Rat Gene 1.0 ST Array 10860483 - -
          Affymetrix Rat Gene 1.0 ST Array 10860494 - -
          Affymetrix Rat Gene 1.0 ST Array 10860492 - -
          Affymetrix Rat Gene 1.0 ST Array 10860487 - -
          Affymetrix Rat Gene 1.0 ST Array 10860488 - -
          Affymetrix Rat Gene 1.0 ST Array 10860496 - -
          Affymetrix Rat Gene 1.0 ST Array 10860491 - -
          Affymetrix Rat Gene 1.0 ST Array 10860482 - -
          Affymetrix Rat Gene 1.0 ST Array 10860498 - -
          Affymetrix Rat Gene 1.0 ST Array 10860493 - -
          Affymetrix Rat Gene 1.1 ST Array 10860481 - -
          Rat Genome 230 2.0 Array 1387723_PM_at - -
          Rat Genome 230 2.0 Array 1387723_at - -
          Affymetrix HT Rat Focus Array 1387723_PM_at - -
          Affymetrix HT Rat Focus Array 1387723_at - -
          Affymetrix Rat Clariom S Assay PSR0400016338.rn.1 TC0400002298.rn.1 RTA TC
          Rat Expression Set 230 1387723_at - -
          Rat Expression Set 230 1387723_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10860481 - -
          Rat Genome U34 Set X95286cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21274306 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375188 - -
          RatRef-12 Expression BeadChip ILMN_64332 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline