Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00437351_m1
          See other WNT6 GE Assays ›
          Gene Symbol:
          Wnt6
          Gene Name:
          wingless-type MMTV integration site family, member 6
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 82033543 - 82047172 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00437351_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Wnt6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108226.1 NP_001101696.1 1 - 2 323 87

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0900000910.rn.1

          Set Membership:

          Amplicon length between 86 and 100 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          branching involved in ureteric bud morphogenesis
          biological_process
          axis specification
          positive regulation of gene expression
          Wnt signaling pathway
          neuron differentiation
          odontogenesis of dentin-containing tooth
          cell fate commitment
          positive regulation of transcription, DNA-templated
          epithelial-mesenchymal cell signaling
          positive regulation of tooth mineralization
          nephron tubule formation
          nephron tubule development
          molecular_function
          frizzled binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P071451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810480 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 363769 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 738926 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874263 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 364404 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840584 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960597 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91506 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 157781 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134460 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44157 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071451 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P071451 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P071451 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P140645 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14498 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10924425 - -
          Affymetrix Rat Gene 1.0 ST Array 10924424 - -
          Affymetrix Rat Gene 1.0 ST Array 10924426 - -
          Affymetrix Rat Gene 1.0 ST Array 10924423 - -
          Affymetrix Rat Gene 1.1 ST Array 10924422 - -
          Rat Genome 230 2.0 Array 1376063_at - -
          Rat Genome 230 2.0 Array 1391914_PM_at - -
          Rat Genome 230 2.0 Array 1391914_at - -
          Rat Genome 230 2.0 Array 1376063_PM_at - -
          Affymetrix HT Rat Focus Array 1391914_at - -
          Affymetrix HT Rat Focus Array 1376063_at - -
          Affymetrix HT Rat Focus Array 1391914_PM_at - -
          Affymetrix HT Rat Focus Array 1376063_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0900007612.rn.1 TC0900000910.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0900007613.rn.1 TC0900000910.rn.1 RTA TC
          Rat Expression Set 230 1376063_PM_at - -
          Rat Expression Set 230 1376063_at - -
          Rat Expression Set 230 1391914_at - -
          Rat Expression Set 230 1391914_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10924422 - -
          Rat Genome U34 Set rc_AI230233_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21865555 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358623 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline