Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00438221_m1
          See other CKMT1B GE Assays ›
          Gene Symbol:
          Ckmt1b
          Gene Name:
          creatine kinase, mitochondrial 1B
          Gene Aliases:
          Ckmt1
          Chromosome Location:
          Chr.3: 113318556 - 113324459 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00438221_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ckmt1b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001012738.1 NP_001012756.1 5 - 6 1078 84
          XM_006234830.2 XP_006234892.1 6 - 7 1034 84
          GenBank mRNA BC091335.1 - 5 - 6 1078 7314893 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0300001560.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ckmt1b

          Panther Classification:

          Molecular Function -

          amino acid kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphorylation
          negative regulation of protein binding
          negative regulation of apoptotic process
          phosphocreatine biosynthetic process
          creatine kinase activity
          ATP binding
          kinase activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1037972 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 16370 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107617 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15546 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037972 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1037972 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1037972 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1037972 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66772 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10839150 - -
          Affymetrix Rat Gene 1.0 ST Array 10839153 - -
          Affymetrix Rat Gene 1.0 ST Array 10839147 - -
          Affymetrix Rat Gene 1.0 ST Array 10839145 - -
          Affymetrix Rat Gene 1.0 ST Array 10839151 - -
          Affymetrix Rat Gene 1.0 ST Array 10839146 - -
          Affymetrix Rat Gene 1.0 ST Array 10839148 - -
          Affymetrix Rat Gene 1.0 ST Array 10839149 - -
          Affymetrix Rat Gene 1.0 ST Array 10839152 - -
          Affymetrix Rat Gene 1.1 ST Array 10839144 - -
          Rat Genome 230 2.0 Array 1390566_a_at - -
          Rat Genome 230 2.0 Array 1390566_PM_a_at - -
          Affymetrix HT Rat Focus Array 1390566_PM_a_at - -
          Affymetrix HT Rat Focus Array 1390566_a_at - -
          Affymetrix Rat Clariom S Assay PSR0300010838.rn.1 TC0300001560.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0300010836.rn.1 TC0300001560.rn.1 RTA TC
          Rat Expression Set 230 1390566_PM_a_at - -
          Rat Expression Set 230 1390566_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10839144 - -
          Rat Genome U34 Set X59737mRNA_at - -
          Rat Genome U34 Set X59737mRNA_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20956902 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-t4kr2:80/100.66.77.141:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0