Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00440891_m1
          See other PRKCD GE Assays ›
          Gene Symbol:
          Prkcd
          Gene Name:
          protein kinase C, delta
          Gene Aliases:
          Pkcd
          Chromosome Location:
          Chr.16: 6655131 - 6675746 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00440891_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prkcd
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133307.1 NP_579841.1 17 - 18 2227 58
          GenBank mRNA AF219629.1 - 17 - 18 2304 58
          BC076505.1 - 16 - 17 2053 7099952 58
          FQ235241.1 - 1 - 2 54 58
          M18330.1 - 17 - 18 2227 58

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600001155.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Protein Kinase 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus Rat Estrogen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prkcd

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          protein phosphorylation
          apoptotic process
          response to oxidative stress
          aging
          response to heat
          response to mechanical stimulus
          response to glucose
          response to organonitrogen compound
          response to organic cyclic compound
          aspartate transport
          immunoglobulin mediated immune response
          peptidyl-threonine phosphorylation
          termination of signal transduction
          negative regulation of actin filament polymerization
          interleukin-10 production
          interleukin-12 production
          cellular response to insulin stimulus
          positive regulation of superoxide anion generation
          collagen metabolic process
          negative regulation of glial cell apoptotic process
          positive regulation of protein dephosphorylation
          intracellular signal transduction
          B cell proliferation
          neutrophil activation
          cellular response to glucose starvation
          response to drug
          response to hydrogen peroxide
          defense response to bacterium
          positive regulation of apoptotic process
          positive regulation of catalytic activity
          response to amino acid
          positive regulation of MAP kinase activity
          negative regulation of MAP kinase activity
          positive regulation of MAPK cascade
          response to ethanol
          positive regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          protein autophosphorylation
          negative regulation of peptidyl-tyrosine phosphorylation
          negative regulation of filopodium assembly
          negative regulation of platelet aggregation
          cellular senescence
          positive regulation of phospholipid scramblase activity
          positive regulation of ceramide biosynthetic process
          positive regulation of glucosylceramide catabolic process
          positive regulation of sphingomyelin catabolic process
          positive regulation of response to DNA damage stimulus
          positive regulation of apoptotic signaling pathway
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          protein binding
          ATP binding
          enzyme activator activity
          enzyme binding
          protein kinase binding
          insulin receptor substrate binding
          metal ion binding
          TIR domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P451071 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595541 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216374 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 69168 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 594906 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 472667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 209562 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591747 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 123500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 248507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 951781 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877141 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 561417 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 507020 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 189551 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83287 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 75016 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P451071 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P451071 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P451071 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P451071 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700920211 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700310435 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46421 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26325 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4634 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92002 - -
          Agilent Rat cDNA Microarray (G4105A) 700310435 - -
          Agilent Rat cDNA Microarray (G4105A) 700920211 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789942 - -
          Affymetrix Rat Gene 1.0 ST Array 10789946 - -
          Affymetrix Rat Gene 1.0 ST Array 10789938 - -
          Affymetrix Rat Gene 1.0 ST Array 10789934 - -
          Affymetrix Rat Gene 1.0 ST Array 10789932 - -
          Affymetrix Rat Gene 1.0 ST Array 10789937 - -
          Affymetrix Rat Gene 1.0 ST Array 10789936 - -
          Affymetrix Rat Gene 1.0 ST Array 10789945 - -
          Affymetrix Rat Gene 1.0 ST Array 10789935 - -
          Affymetrix Rat Gene 1.0 ST Array 10789933 - -
          Affymetrix Rat Gene 1.0 ST Array 10789944 - -
          Affymetrix Rat Gene 1.0 ST Array 10789931 - -
          Affymetrix Rat Gene 1.0 ST Array 10789947 - -
          Affymetrix Rat Gene 1.0 ST Array 10789941 - -
          Affymetrix Rat Gene 1.0 ST Array 10789943 - -
          Affymetrix Rat Gene 1.0 ST Array 10789948 - -
          Affymetrix Rat Gene 1.0 ST Array 10789930 - -
          Affymetrix Rat Gene 1.0 ST Array 10789940 - -
          Affymetrix Rat Gene 1.0 ST Array 10789949 - -
          Affymetrix Rat Gene 1.1 ST Array 10789929 - -
          Rat Genome 230 2.0 Array 1387114_at - -
          Rat Genome 230 2.0 Array 1387114_PM_at - -
          Affymetrix HT Rat Focus Array 1387114_PM_at - -
          Affymetrix HT Rat Focus Array 1387114_at - -
          Affymetrix Rat Clariom S Assay PSR1600008516.rn.1 TC1600001155.rn.1 RTA TC
          Rat Expression Set 230 1387114_at - -
          Rat Expression Set 230 1387114_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789929 - -
          Rat Genome U34 Set M18330_at - -
          Rat Genome U34 Set M18330_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21409579 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1360400 - -
          RatRef-12 Expression BeadChip ILMN_58185 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline