Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00440932_m1
          See other PRRX1 GE Assays ›
          Gene Symbol:
          Prrx1
          Gene Name:
          paired related homeobox 1
          Gene Aliases:
          Pmx1, Prx-1
          Chromosome Location:
          Chr.13: 81147038 - 81215560 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00440932_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prrx1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_153821.1 NP_722543.1 2 - 3 741 65
          XM_006250151.2 XP_006250213.1 2 - 3 1479 65
          GenBank mRNA AY730764.1 - 2 - 3 465 65
          S82911.1 - 2 - 3 741 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1300001786.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prrx1

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          positive regulation of mesenchymal cell proliferation
          embryonic limb morphogenesis
          inner ear morphogenesis
          middle ear morphogenesis
          positive regulation of smoothened signaling pathway
          positive regulation of transcription from RNA polymerase II promoter
          neuron fate determination
          embryonic cranial skeleton morphogenesis
          embryonic skeletal system morphogenesis
          artery morphogenesis
          cartilage development
          palate development
          regulation of neuron projection regeneration
          neuronal stem cell population maintenance
          RNA polymerase II transcription coactivator activity
          DNA binding
          sequence-specific DNA binding
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P479890 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P093507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 615254 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 798624 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 370277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 434113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 696619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814219 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 423617 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 406136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 11187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 324920 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 574615 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 463607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 297968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61696 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 727443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34798 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 656805 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 791271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 235667 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 2048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 726931 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 719814 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 140656 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 437438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36039 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 919543 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 546015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 687740 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 266567 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 408556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 736573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714405 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272780 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 38110 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 7437 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74723 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15273 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38794 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 15954 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 111803 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P479890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093507 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P479890 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P093507 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P093507 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P479890 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P479890 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700537335 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92024 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 67638 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 48639 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72661 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26288 - -
          Agilent Rat cDNA Microarray (G4105A) 700537335 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10769427 - -
          Affymetrix Rat Gene 1.0 ST Array 10769425 - -
          Affymetrix Rat Gene 1.0 ST Array 10769428 - -
          Affymetrix Rat Gene 1.0 ST Array 10769424 - -
          Affymetrix Rat Gene 1.0 ST Array 10769429 - -
          Affymetrix Rat Gene 1.0 ST Array 10769423 - -
          Affymetrix Rat Gene 1.0 ST Array 10769426 - -
          Affymetrix Rat Gene 1.1 ST Array 10769422 - -
          Rat Genome 230 2.0 Array 1384234_PM_at - -
          Rat Genome 230 2.0 Array 1384234_at - -
          Rat Genome 230 2.0 Array 1375485_PM_at - -
          Rat Genome 230 2.0 Array 1375485_at - -
          Affymetrix HT Rat Focus Array 1375485_PM_at - -
          Affymetrix HT Rat Focus Array 1384234_at - -
          Affymetrix HT Rat Focus Array 1384234_PM_at - -
          Affymetrix HT Rat Focus Array 1375485_at - -
          Affymetrix Rat Clariom S Assay PSR1300012682.rn.1 TC1300001786.rn.1 RTA TC
          Rat Expression Set 230 1375485_at - -
          Rat Expression Set 230 1375485_PM_at - -
          Rat Expression Set 230 1384234_PM_at - -
          Rat Expression Set 230 1384234_at - -
          Affymetrix Rat Gene 1.0 ST Array 10769422 - -
          Rat Genome U34 Set S82911_at - -
          Rat Genome U34 Set rc_AA957064_g_at - -
          Rat Genome U34 Set rc_AA957064_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21740081 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368514 - -
          RatRef-12 Expression BeadChip ILMN_63084 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline