Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00457825_m1
          See other CACNA2D2 GE Assays ›
          Gene Symbol:
          Cacna2d2
          Gene Name:
          calcium channel, voltage-dependent, alpha 2/delta subunit 2
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 116154462 - 116285643 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00457825_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cacna2d2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_175592.2 NP_783182.1 1 - 2 218 84
          XM_006243734.2 XP_006243796.1 1 - 2 492 84
          XM_006243735.2 XP_006243797.1 1 - 2 492 84
          XM_006243736.2 XP_006243798.1 1 - 2 492 84
          XM_006243737.2 XP_006243799.1 1 - 2 492 84
          GenBank mRNA AF486277.1 - 1 - 2 218 84

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800001501.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cacna2d2

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          calcium ion transport
          neuromuscular junction development
          regulation of ion transmembrane transport
          regulation of multicellular organism growth
          positive regulation of organ growth
          muscle fiber development
          rhythmic synaptic transmission
          calcium ion transmembrane transport
          voltage-gated calcium channel activity
          calcium channel regulator activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P137630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 396579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 869077 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 773111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 547503 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 764385 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 151165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 383421 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 892204 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 571092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 834628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127288 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 215350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 954640 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 823959 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 626629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575881 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 652373 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 874166 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705829 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147726 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 61396 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 647699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 87709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 592434 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 782190 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45386 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 105099 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 274016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 357045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 552128 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 554234 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636560 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 27231 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20295 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741964 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 907551 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 102256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 135416 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655936 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359938 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 916962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469143 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335947 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 362737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450534 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 958501 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373536 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 99033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 897527 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 537011 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 118148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 776820 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 384658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 832524 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 488041 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 474791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 640790 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844554 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915983 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 703731 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 740644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262067 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487985 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 237989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 889644 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 742428 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 165953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 559786 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 595224 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 585165 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 400973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 649275 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 520845 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 714925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 661118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 806079 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 36758 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361901 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724008 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 908824 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32688 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 78306 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 19479 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 115921 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 69798 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 84022 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 74415 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 29393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124741 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 81052 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23951 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124062 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 58293 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 44124 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137630 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P137630 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P137630 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P217286 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 3452 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35753 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 33548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 60064 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 99630 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72529 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25511 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 61284 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10912918 - -
          Affymetrix Rat Gene 1.0 ST Array 10912954 - -
          Affymetrix Rat Gene 1.0 ST Array 10912955 - -
          Affymetrix Rat Gene 1.0 ST Array 10912929 - -
          Affymetrix Rat Gene 1.0 ST Array 10912927 - -
          Affymetrix Rat Gene 1.0 ST Array 10912938 - -
          Affymetrix Rat Gene 1.0 ST Array 10912950 - -
          Affymetrix Rat Gene 1.0 ST Array 10912946 - -
          Affymetrix Rat Gene 1.0 ST Array 10912928 - -
          Affymetrix Rat Gene 1.0 ST Array 10912947 - -
          Affymetrix Rat Gene 1.0 ST Array 10912921 - -
          Affymetrix Rat Gene 1.0 ST Array 10912925 - -
          Affymetrix Rat Gene 1.0 ST Array 10912922 - -
          Affymetrix Rat Gene 1.0 ST Array 10912924 - -
          Affymetrix Rat Gene 1.0 ST Array 10912948 - -
          Affymetrix Rat Gene 1.0 ST Array 10912944 - -
          Affymetrix Rat Gene 1.0 ST Array 10912934 - -
          Affymetrix Rat Gene 1.0 ST Array 10912943 - -
          Affymetrix Rat Gene 1.0 ST Array 10912920 - -
          Affymetrix Rat Gene 1.0 ST Array 10912940 - -
          Affymetrix Rat Gene 1.0 ST Array 10912919 - -
          Affymetrix Rat Gene 1.0 ST Array 10912923 - -
          Affymetrix Rat Gene 1.0 ST Array 10912951 - -
          Affymetrix Rat Gene 1.0 ST Array 10912932 - -
          Affymetrix Rat Gene 1.0 ST Array 10912926 - -
          Affymetrix Rat Gene 1.0 ST Array 10912942 - -
          Affymetrix Rat Gene 1.0 ST Array 10912939 - -
          Affymetrix Rat Gene 1.0 ST Array 10912953 - -
          Affymetrix Rat Gene 1.0 ST Array 10912949 - -
          Affymetrix Rat Gene 1.0 ST Array 10912933 - -
          Affymetrix Rat Gene 1.0 ST Array 10912930 - -
          Affymetrix Rat Gene 1.0 ST Array 10912936 - -
          Affymetrix Rat Gene 1.0 ST Array 10912935 - -
          Affymetrix Rat Gene 1.0 ST Array 10912941 - -
          Affymetrix Rat Gene 1.0 ST Array 10912937 - -
          Affymetrix Rat Gene 1.0 ST Array 10912945 - -
          Affymetrix Rat Gene 1.0 ST Array 10912952 - -
          Affymetrix Rat Gene 1.0 ST Array 10912931 - -
          Affymetrix Rat Gene 1.1 ST Array 10912917 - -
          Affymetrix Rat Clariom S Assay PSR0800010957.rn.1 TC0800001501.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800010956.rn.1 TC0800001501.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10912917 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21781539 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362200 - -
          RatRef-12 Expression BeadChip ILMN_55233 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline