Search
Search

Slc25a45| Gene Symbol: Slc25a45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271179.1 | NP_001258108.1 | 5 - 6 | 593 | 58 | |
| XM_008760179.1 | XP_008758401.1 | 5 - 6 | 893 | 58 | ||
| GenBank mRNA | FM090288.1 | - | 1 - 2 | 165 | 58 | |
Set Membership: |
|