Search
Search

Slc25a1| Gene Symbol: Slc25a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017307.3 | NP_059003.1 | 3 - 4 | 414 | 66 | |
| GenBank mRNA | BC100077.1 | - | 3 - 4 | 398 | 7126394 | 66 |
Set Membership: |
|