Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00469666_m1
          See other FXYD7 GE Assays ›
          Gene Symbol:
          Fxyd7
          Gene Name:
          FXYD domain-containing ion transport regulator 7
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 89474601 - 89484068 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00469666_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fxyd7
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_022008.1 NP_071291.1 5 - 6 305 74
          XM_006228843.2 XP_006228905.1 5 - 6 376 74
          GenBank mRNA AW914994.1 - 5 - 6 305 74

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100005181.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fxyd7

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          regulation of ion transport
          ion channel activity
          ATPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P1056589 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 80773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 436533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85992 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577544 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 578678 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 582036 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 912099 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118644 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056589 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P1056589 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P1056589 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P1056589 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 77473 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10720870 - -
          Affymetrix Rat Gene 1.0 ST Array 10720876 - -
          Affymetrix Rat Gene 1.0 ST Array 10720871 - -
          Affymetrix Rat Gene 1.0 ST Array 10720874 - -
          Affymetrix Rat Gene 1.0 ST Array 10720873 - -
          Affymetrix Rat Gene 1.0 ST Array 10720872 - -
          Affymetrix Rat Gene 1.0 ST Array 10720877 - -
          Affymetrix Rat Gene 1.0 ST Array 10720875 - -
          Affymetrix Rat Gene 1.1 ST Array 10720869 - -
          Rat Genome 230 2.0 Array 1368696_PM_at - -
          Rat Genome 230 2.0 Array 1368696_at - -
          Affymetrix HT Rat Focus Array 1368696_PM_at - -
          Affymetrix HT Rat Focus Array 1368696_at - -
          Affymetrix Rat Clariom S Assay PSR0100041810.rn.1 TC0100005181.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100041811.rn.1 TC0100005181.rn.1 RTA TC
          Rat Expression Set 230 1368696_PM_at - -
          Rat Expression Set 230 1368696_at - -
          Affymetrix Rat Gene 1.0 ST Array 10720869 - -
          Rat Genome U34 Set rc_AI028815_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21609234 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356198 - -
          RatRef-12 Expression BeadChip ILMN_65540 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7b6576f7bc-d59m7:80/100.66.79.221:80.
          git-commit: 6d309ebe7a31b2ceadeb87f43b473eebab110cf3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-2026.03.11-1.0