Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00469699_g1
          See other ZFAND6 GE Assays ›
          Gene Symbol:
          Zfand6
          Gene Name:
          zinc finger, AN1-type domain 6
          Gene Aliases:
          Za20d3, zgc:101121
          Chromosome Location:
          Chr.1: 146745850 - 146823762 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00469699_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Zfand6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001007630.1 NP_001007631.1 6 - 7 631 65
          XM_006229499.2 XP_006229561.1 8 - 9 936 65
          XM_006229500.2 XP_006229562.1 6 - 7 616 65
          XM_006229502.2 XP_006229564.1 7 - 8 759 65
          XM_006229504.1 XP_006229566.1 7 - 8 770 65
          XM_006229505.2 XP_006229567.1 6 - 7 624 65
          XM_006229506.2 XP_006229568.1 6 - 7 621 65
          XM_006229507.2 XP_006229569.1 5 - 6 582 65
          XM_006229508.2 XP_006229570.1 5 - 6 577 65
          XM_008759611.1 XP_008757833.1 6 - 7 2161 65
          GenBank mRNA BC076394.1 - 6 - 7 631 7107784 65

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Zfand6

          Gene Ontology Categories:

          Function(s) Process(es)

          protein targeting to peroxisome
          apoptotic process
          signal transduction
          negative regulation of apoptotic process
          regulation of I-kappaB kinase/NF-kappaB signaling
          cellular response to tumor necrosis factor
          DNA binding
          zinc ion binding
          polyubiquitin binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P370294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 733582 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 888789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 245602 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 758253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862965 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575029 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 695292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 924096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 287244 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 489185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 601594 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 838826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840302 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 830713 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577249 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 375487 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 282188 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628307 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910199 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 654430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195038 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 199630 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 767718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 902750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 342073 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 182154 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156496 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 196031 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159944 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967240 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 947540 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502481 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 34711 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 8449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 677748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 804720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 388807 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 896057 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 533351 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 597718 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 181859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 322699 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 539334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439320 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 452068 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799243 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 326201 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57600 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37178 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 51672 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151163 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 154710 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13124 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174042 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 148602 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13969 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P370294 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P370294 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P370294 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P370294 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23152 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 35873 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57485 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 93923 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 36828 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 47574 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10723463 - -
          Affymetrix Rat Gene 1.0 ST Array 10723461 - -
          Affymetrix Rat Gene 1.0 ST Array 10723462 - -
          Affymetrix Rat Gene 1.0 ST Array 10723458 - -
          Affymetrix Rat Gene 1.0 ST Array 10723459 - -
          Affymetrix Rat Gene 1.0 ST Array 10723460 - -
          Affymetrix Rat Gene 1.1 ST Array 10723457 - -
          Rat Genome 230 2.0 Array 1371464_PM_at - -
          Rat Genome 230 2.0 Array 1371464_at - -
          Affymetrix HT Rat Focus Array 1371464_PM_at - -
          Affymetrix HT Rat Focus Array 1371464_at - -
          Affymetrix Rat Clariom S Assay PSR0100066748.rn.1 TC0100008125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100066747.rn.1 TC0100008125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100066749.rn.1 TC0100008125.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100066752.rn.1 TC0100008125.rn.1 RTA TC
          Rat Expression Set 230 1371464_at - -
          Rat Expression Set 230 1371464_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10723457 - -
          Rat Genome U34 Set rc_AA892376_at - -
          Rat Genome U34 Set AA801153_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21067553 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370487 - -
          RatRef-12 Expression BeadChip ILMN_63277 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-l697b:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline