Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00484683_m1
          See other GATA3 GE Assays ›
          Gene Symbol:
          Gata3
          Gene Name:
          GATA binding protein 3
          Gene Aliases:
          -
          Chromosome Location:
          Chr.17: 72426950 - 72452043 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00484683_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gata3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_133293.1 NP_579827.1 1 - 2 240 57
          XM_006254253.2 XP_006254315.1 2 - 3 1006 57
          XM_008771893.1 XP_008770115.1 2 - 3 950 57
          XM_008771894.1 XP_008770116.1 2 - 3 2540 57
          GenBank mRNA AB000217.1 - 1 - 2 451 57
          AY024364.1 - 1 - 2 240 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700000947.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gata3

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          cell morphogenesis
          in utero embryonic development
          cell fate determination
          neuron migration
          cell activation
          type IV hypersensitivity
          positive regulation of cytokine production
          kidney development
          mesonephros development
          lens development in camera-type eye
          pro-T cell differentiation
          aortic valve morphogenesis
          cardiac right ventricle morphogenesis
          ventricular septum development
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          humoral immune response
          signal transduction
          nervous system development
          axon guidance
          negative regulation of cell proliferation
          male gonad development
          response to virus
          post-embryonic development
          positive regulation of signal transduction
          response to gamma radiation
          positive regulation of endothelial cell migration
          negative regulation of gene expression
          regulation of neuron projection development
          phosphatidylinositol 3-kinase signaling
          neuron differentiation
          T cell differentiation
          erythrocyte differentiation
          TOR signaling
          negative regulation of interferon-gamma production
          negative regulation of interleukin-2 production
          positive regulation of interleukin-13 production
          positive regulation of interleukin-4 production
          positive regulation of interleukin-5 production
          T cell differentiation in thymus
          negative regulation of mammary gland epithelial cell proliferation
          embryonic hemopoiesis
          cellular response to interferon-alpha
          ureter maturation
          parathyroid hormone secretion
          regulation of cytokine biosynthetic process
          norepinephrine biosynthetic process
          inner ear morphogenesis
          response to drug
          regulation of CD4-positive, alpha-beta T cell differentiation
          regulation of neuron apoptotic process
          ear development
          response to estrogen
          thymic T cell selection
          T-helper 2 cell differentiation
          response to ethanol
          positive regulation of T cell differentiation
          positive regulation of cell differentiation
          negative regulation of fat cell differentiation
          negative regulation of cell cycle
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          sympathetic nervous system development
          thymus development
          embryonic organ development
          developmental growth
          anatomical structure formation involved in morphogenesis
          homeostasis of number of cells
          negative regulation of inflammatory response
          T cell receptor signaling pathway
          regulation of histone H3-K4 methylation
          positive regulation of protein kinase B signaling
          parathyroid gland development
          pharyngeal system development
          uterus development
          mesenchymal to epithelial transition
          mast cell differentiation
          ureteric bud formation
          regulation of histone H3-K27 methylation
          canonical Wnt signaling pathway involved in metanephric kidney development
          cellular response to cytokine stimulus
          cellular response to interleukin-4
          cellular response to tumor necrosis factor
          otic vesicle development
          cellular response to BMP stimulus
          renal system development
          positive regulation of ureteric bud formation
          nephric duct morphogenesis
          nephric duct formation
          regulation of nephron tubule epithelial cell differentiation
          interleukin-4 secretion
          interferon-gamma secretion
          lymphocyte migration
          regulation of establishment of cell polarity
          negative regulation of cell motility
          negative regulation of endothelial cell apoptotic process
          negative regulation of cell proliferation involved in mesonephros development
          positive regulation of thyroid hormone generation
          positive regulation of interleukin-5 secretion
          positive regulation of interleukin-13 secretion
          positive regulation of transcription regulatory region DNA binding
          regulation of cellular response to X-ray
          negative regulation of fibroblast growth factor receptor signaling pathway involved in ureteric bud formation
          negative regulation of glial cell-derived neurotrophic factor receptor signaling pathway involved in ureteric bud formation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter sequence-specific DNA binding
          core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
          enhancer sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription coactivator activity
          interleukin-2 receptor binding
          transcription factor binding
          zinc ion binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          protein dimerization activity
          E-box binding
          HMG box domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P117700 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 49626 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 459535 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891449 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 424818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 422491 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692561 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 502704 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 632251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 217372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 650102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271896 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103146 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 439152 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 658498 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 138363 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163381 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117700 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P117700 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P117700 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P383310 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 14174 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15254 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24768 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796222 - -
          Affymetrix Rat Gene 1.0 ST Array 10796223 - -
          Affymetrix Rat Gene 1.0 ST Array 10796225 - -
          Affymetrix Rat Gene 1.0 ST Array 10796224 - -
          Affymetrix Rat Gene 1.0 ST Array 10796221 - -
          Affymetrix Rat Gene 1.1 ST Array 10796220 - -
          Rat Genome 230 2.0 Array 1369525_at - -
          Rat Genome 230 2.0 Array 1382163_PM_at - -
          Rat Genome 230 2.0 Array 1382163_at - -
          Rat Genome 230 2.0 Array 1369525_PM_at - -
          Affymetrix HT Rat Focus Array 1382163_PM_at - -
          Affymetrix HT Rat Focus Array 1382163_at - -
          Affymetrix HT Rat Focus Array 1369525_at - -
          Affymetrix HT Rat Focus Array 1369525_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1700006579.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006584.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006586.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006581.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006576.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006575.rn.1 TC1700000947.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700006577.rn.1 TC1700000947.rn.1 RTA TC
          Rat Expression Set 230 1369525_at - -
          Rat Expression Set 230 1369525_PM_at - -
          Rat Expression Set 230 1382163_at - -
          Rat Expression Set 230 1382163_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10796220 - -
          Rat Genome U34 Set AB000217mRNA_at - -
          Rat Genome U34 Set rc_AI230345_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20859458 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371339 - -
          RatRef-12 Expression BeadChip ILMN_50466 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline