Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00485207_m1
          See other MEIS3 GE Assays ›
          Gene Symbol:
          Meis3
          Gene Name:
          Meis homeobox 3
          Gene Aliases:
          Mrg2
          Chromosome Location:
          Chr.1: 78068337 - 78079698 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00485207_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Meis3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001108472.1 NP_001101942.1 5 - 6 597 70
          XM_006228346.2 XP_006228408.1 4 - 5 658 70
          XM_006228347.2 XP_006228409.1 5 - 6 1191 70
          XM_006228348.2 XP_006228410.1 4 - 5 658 70
          XM_006228349.2 XP_006228411.1 4 - 5 658 70
          XM_006228350.2 XP_006228412.1 4 - 5 658 70
          XM_006228351.2 XP_006228413.1 4 - 5 658 70
          XM_008758950.1 XP_008757172.1 4 - 5 658 70
          GenBank mRNA BC160854.1 - 4 - 5 558 8364874 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100000924.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of protein kinase B signaling
          negative regulation of apoptotic signaling pathway
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          DNA binding
          chromatin binding
          sequence-specific DNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P285357 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660045 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73378 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 674003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 824645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412139 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 228214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 788348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743315 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432533 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 123886 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 6178 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19545 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P19545 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P19545 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P19545 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 57997 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10704466 - -
          Affymetrix Rat Gene 1.0 ST Array 10704463 - -
          Affymetrix Rat Gene 1.0 ST Array 10704453 - -
          Affymetrix Rat Gene 1.0 ST Array 10704460 - -
          Affymetrix Rat Gene 1.0 ST Array 10704465 - -
          Affymetrix Rat Gene 1.0 ST Array 10704454 - -
          Affymetrix Rat Gene 1.0 ST Array 10704458 - -
          Affymetrix Rat Gene 1.0 ST Array 10704456 - -
          Affymetrix Rat Gene 1.0 ST Array 10704455 - -
          Affymetrix Rat Gene 1.0 ST Array 10704464 - -
          Affymetrix Rat Gene 1.0 ST Array 10704457 - -
          Affymetrix Rat Gene 1.0 ST Array 10704462 - -
          Affymetrix Rat Gene 1.0 ST Array 10704459 - -
          Affymetrix Rat Gene 1.1 ST Array 10704452 - -
          Rat Genome 230 2.0 Array 1372800_PM_at - -
          Rat Genome 230 2.0 Array 1372800_at - -
          Affymetrix HT Rat Focus Array 1372800_at - -
          Affymetrix HT Rat Focus Array 1372800_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100006336.rn.1 TC0100000924.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100006335.rn.1 TC0100000924.rn.1 RTA TC
          Rat Expression Set 230 1372800_at - -
          Rat Expression Set 230 1372800_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10704452 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20776766 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1364323 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline