Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00487218_m1
          See other CYP1A1 GE Assays ›
          Gene Symbol:
          Cyp1a1
          Gene Name:
          cytochrome P450, family 1, subfamily a, polypeptide 1
          Gene Aliases:
          AHH, AHRR, CP11, CYP1, Cyp45c, Cypc45c, P-450MC, P1-450, P450-C, P450DX
          Chromosome Location:
          Chr.8: 62472087 - 62478122 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00487218_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cyp1a1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012540.2 NP_036672.2 2 - 3 906 120
          XM_006243150.2 XP_006243212.1 2 - 3 903 120
          GenBank mRNA X00469.1 - 2 - 3 906 120
          X17160.1 - 916 120

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0800000802.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 P450 Detect Genomic DNA Recommended Assay Rattus norvegicus Rat Estrogen Panel Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cyp1a1

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          liver development
          porphyrin-containing compound metabolic process
          aging
          cell proliferation
          amine metabolic process
          toxin metabolic process
          response to wounding
          response to virus
          response to nematode
          response to herbicide
          response to toxic substance
          embryo development ending in birth or egg hatching
          coumarin metabolic process
          flavonoid metabolic process
          response to organic substance
          response to iron(III) ion
          response to organic cyclic compound
          insecticide metabolic process
          drug metabolic process
          dibenzo-p-dioxin metabolic process
          dibenzo-p-dioxin catabolic process
          response to food
          response to lipopolysaccharide
          developmental process
          response to vitamin A
          response to immobilization stress
          response to drug
          9-cis-retinoic acid biosynthetic process
          camera-type eye development
          heterocycle metabolic process
          response to antibiotic
          response to arsenic-containing substance
          digestive tract development
          hydrogen peroxide biosynthetic process
          response to hyperoxia
          oxidation-reduction process
          maternal process involved in parturition
          hepatocyte differentiation
          demethylation
          cellular response to organic cyclic compound
          positive regulation of G1/S transition of mitotic cell cycle
          catalytic activity
          monooxygenase activity
          iron ion binding
          protein binding
          steroid hydroxylase activity
          oxidoreductase activity
          oxidoreductase activity, acting on diphenols and related substances as donors
          flavonoid 3'-monooxygenase activity
          oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced flavin or flavoprotein as one donor, and incorporation of one atom of oxygen
          enzyme binding
          heme binding
          demethylase activity
          aromatase activity
          vitamin D 24-hydroxylase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Clariom S Assay PSR0800005499.rn.1 TC0800000802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800005502.rn.1 TC0800000802.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0800005501.rn.1 TC0800000802.rn.1 RTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline