Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00488446_m1
          See other STAM GE Assays ›
          Gene Symbol:
          Stam
          Gene Name:
          signal transducing adaptor molecule (SH3 domain and ITAM motif) 1
          Gene Aliases:
          RGD1564499
          Chromosome Location:
          Chr.17: 81209867 - 81255808 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00488446_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stam
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109121.2 NP_001102591.1 1 - 2 157 63
          GenBank mRNA BC168933.1 - 1 - 2 157 7106331 63
          FQ221736.1 - 1 - 2 234 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1700001051.rn.1

          Set Membership:

          Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          intracellular protein transport
          positive regulation of exosomal secretion
          regulation of extracellular exosome assembly
          protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P128564 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P986963 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 725872 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402384 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302581 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 560910 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 659803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 277870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 281331 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 127614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821974 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 252804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 540411 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195402 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 671701 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 234042 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 98578 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345078 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 562748 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 417789 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 238134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 40236 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660868 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 536049 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884147 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 355081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 113492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 469871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 285843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 973702 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 730737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971222 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159713 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 33455 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 99185 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118123 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 97841 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128564 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P986963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P986963 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P128564 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P128564 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P986963 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P128564 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P986963 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 76356 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10796462 - -
          Affymetrix Rat Gene 1.0 ST Array 10796467 - -
          Affymetrix Rat Gene 1.0 ST Array 10796456 - -
          Affymetrix Rat Gene 1.0 ST Array 10796465 - -
          Affymetrix Rat Gene 1.0 ST Array 10796460 - -
          Affymetrix Rat Gene 1.0 ST Array 10796457 - -
          Affymetrix Rat Gene 1.0 ST Array 10796459 - -
          Affymetrix Rat Gene 1.0 ST Array 10796458 - -
          Affymetrix Rat Gene 1.0 ST Array 10796464 - -
          Affymetrix Rat Gene 1.0 ST Array 10796466 - -
          Affymetrix Rat Gene 1.0 ST Array 10796461 - -
          Affymetrix Rat Gene 1.0 ST Array 10796463 - -
          Affymetrix Rat Gene 1.1 ST Array 10796455 - -
          Affymetrix Rat Clariom S Assay PSR1700007195.rn.1 TC1700001051.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1700007192.rn.1 TC1700001051.rn.1 RTA TC
          Affymetrix Rat Gene 1.0 ST Array 10796455 - -
          Rat Genome U34 Set rc_AI175294_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22300539 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline