Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00492098_g1
          See other DDIT3 GE Assays ›
          Gene Symbol:
          Ddit3
          Gene Name:
          DNA-damage inducible transcript 3
          Gene Aliases:
          CHOP, CHOP-10, Chop10, Gadd153
          Chromosome Location:
          Chr.7: 70578545 - 70585074 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00492098_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ddit3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001109986.1 NP_001103456.1 3 - 4 305 92
          NM_024134.2 NP_077048.2 2 - 3 257 92
          XM_006241444.2 XP_006241506.1 4 - 5 1483 92
          XM_006241445.2 XP_006241507.1 3 - 4 1435 92
          GenBank mRNA AW916370.1 - 3 - 4 305 92
          BC100664.1 - 3 - 4 268 7932840 92
          BF282473.1 - 1 - 2 92
          CF977695.1 - 2 - 3 260 92
          U30186.1 - 2 - 3 249 92
          U36994.1 - 2 - 3 227 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700000893.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ddit3

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          blood vessel maturation
          response to amphetamine
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          apoptotic process
          response to oxidative stress
          ER overload response
          response to unfolded protein
          cell cycle arrest
          aging
          response to nutrient
          Wnt signaling pathway
          endoplasmic reticulum unfolded protein response
          positive regulation of interleukin-8 production
          negative regulation of CREB transcription factor activity
          response to endoplasmic reticulum stress
          response to drug
          response to hydrogen peroxide
          response to starvation
          mRNA transcription from RNA polymerase II promoter
          positive regulation of apoptotic process
          proteasome-mediated ubiquitin-dependent protein catabolic process
          negative regulation of DNA binding
          negative regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of neuron apoptotic process
          regulation of transcription involved in anterior/posterior axis specification
          cell redox homeostasis
          negative regulation of fat cell differentiation
          negative regulation of myoblast differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          embryonic organ development
          release of sequestered calcium ion into cytosol
          negative regulation of protein kinase B signaling
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          negative regulation of canonical Wnt signaling pathway
          negative regulation of RNA polymerase II regulatory region sequence-specific DNA binding
          positive regulation of transcription from RNA polymerase II promoter in response to endoplasmic reticulum stress
          negative regulation of determination of dorsal identity
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          transcription factor binding
          protein homodimerization activity
          leucine zipper domain binding
          sequence-specific DNA binding
          protein heterodimerization activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P059495 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 460327 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 178694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 808759 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059495 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P059495 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P059495 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P445344 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700180249 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39750 - -
          Agilent Rat cDNA Microarray (G4105A) 700180249 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10895864 - -
          Affymetrix Rat Gene 1.0 ST Array 10895862 - -
          Affymetrix Rat Gene 1.0 ST Array 10895865 - -
          Affymetrix Rat Gene 1.0 ST Array 10895863 - -
          Affymetrix Rat Gene 1.1 ST Array 10895861 - -
          Rat Genome 230 2.0 Array 1369590_PM_a_at - -
          Rat Genome 230 2.0 Array 1369590_a_at - -
          Affymetrix HT Rat Focus Array 1369590_PM_a_at - -
          Affymetrix HT Rat Focus Array 1369590_a_at - -
          Affymetrix Rat Clariom S Assay PSR0700006739.rn.1 TC0700000893.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700006738.rn.1 TC0700000893.rn.1 RTA TC
          Rat Expression Set 230 1369590_a_at - -
          Rat Expression Set 230 1369590_PM_a_at - -
          Affymetrix Rat Gene 1.0 ST Array 10895861 - -
          Rat Genome U34 Set U30186_at - -
          Rat Genome U34 Set AA686470_i_at - -
          Rat Genome U34 Set AA686470_s_at - -
          Rat Genome U34 Set U30186_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21564969 - -
          ABI Rat Genome Survey Microarray 21924040 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349910 - -
          RatRef-12 Expression BeadChip ILMN_69830 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline