Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00498081_g1
          See other RNF138 GE Assays ›
          Gene Symbol:
          Rnf138
          Gene Name:
          ring finger protein 138, E3 ubiquitin protein ligase
          Gene Aliases:
          Rsd4, Trif
          Chromosome Location:
          Chr.18: 15146925 - 15167361 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00498081_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rnf138
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_053588.2 NP_446040.1 500 115
          GenBank mRNA AF315468.1 - 461 115
          BC061821.1 - 500 5599087 115
          FQ220857.1 - 796 115

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rnf138

          Panther Classification:

          Molecular Function -

          ubiquitin-protein ligase

          Gene Ontology Categories:

          Function(s) Process(es)

          Wnt signaling pathway
          protein ubiquitination
          zinc ion binding
          ligase activity
          protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P467928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 73239 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 915962 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 875908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402462 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862804 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 748185 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 796003 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 195096 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96426 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 503209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 588162 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273734 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 792754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 871607 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 185010 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 399924 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 263997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515219 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 28684 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 168199 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 94972 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 175934 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467928 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P467928 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P467928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P467928 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P312217 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32344 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 9467 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10800449 - -
          Affymetrix Rat Gene 1.0 ST Array 10800450 - -
          Affymetrix Rat Gene 1.0 ST Array 10800444 - -
          Affymetrix Rat Gene 1.0 ST Array 10800443 - -
          Affymetrix Rat Gene 1.0 ST Array 10800447 - -
          Affymetrix Rat Gene 1.0 ST Array 10800446 - -
          Affymetrix Rat Gene 1.0 ST Array 10800445 - -
          Affymetrix Rat Gene 1.1 ST Array 10800442 - -
          Rat Genome 230 2.0 Array 1387201_PM_at - -
          Rat Genome 230 2.0 Array 1389258_at - -
          Rat Genome 230 2.0 Array 1389258_PM_at - -
          Rat Genome 230 2.0 Array 1387201_at - -
          Rat Genome 230 2.0 Array 1382379_PM_at - -
          Rat Genome 230 2.0 Array 1382379_at - -
          Affymetrix HT Rat Focus Array 1387201_at - -
          Affymetrix HT Rat Focus Array 1382379_PM_at - -
          Affymetrix HT Rat Focus Array 1387201_PM_at - -
          Affymetrix HT Rat Focus Array 1389258_at - -
          Affymetrix HT Rat Focus Array 1382379_at - -
          Affymetrix HT Rat Focus Array 1389258_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1800007860.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007854.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007853.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007855.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007858.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007856.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007865.rn.1 TC1800001106.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1800007859.rn.1 TC1800001106.rn.1 RTA TC
          Rat Expression Set 230 1389258_at - -
          Rat Expression Set 230 1389258_PM_at - -
          Rat Expression Set 230 1387201_at - -
          Rat Expression Set 230 1387201_PM_at - -
          Rat Expression Set 230 1382379_PM_at - -
          Rat Expression Set 230 1382379_at - -
          Affymetrix Rat Gene 1.0 ST Array 10800442 - -
          Rat Genome U34 Set rc_AA851754_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22169692 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371446 - -
          RatRef-12 Expression BeadChip ILMN_52424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline