Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00502344_m1
          See other STARD10 GE Assays ›
          Gene Symbol:
          Stard10
          Gene Name:
          StAR-related lipid transfer (START) domain containing 10
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 166428779 - 166458413 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00502344_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Stard10
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001013069.1 NP_001013087.1 4 - 5 664 80
          XM_006229850.2 XP_006229912.1 5 - 6 787 80
          XM_006229851.2 XP_006229913.1 4 - 5 726 80
          GenBank mRNA BC091411.1 - 4 - 5 664 7325640 80

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002165.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Stard10

          Gene Ontology Categories:

          Function(s) Process(es)

          bile acid secretion
          positive regulation of peroxisome proliferator activated receptor signaling pathway
          lipid binding

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P105156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 667117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 446448 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 505568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 268124 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644683 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 771156 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 176047 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 859398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 498870 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 873015 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378777 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 905818 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 14836 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 64007 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 138981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 841614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 681308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 337909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207748 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 159073 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 36255 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142548 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105156 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P105156 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P105156 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P105156 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 5205 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 92797 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10709159 - -
          Affymetrix Rat Gene 1.0 ST Array 10709163 - -
          Affymetrix Rat Gene 1.0 ST Array 10709165 - -
          Affymetrix Rat Gene 1.0 ST Array 10709160 - -
          Affymetrix Rat Gene 1.0 ST Array 10709161 - -
          Affymetrix Rat Gene 1.0 ST Array 10709162 - -
          Affymetrix Rat Gene 1.0 ST Array 10709164 - -
          Affymetrix Rat Gene 1.1 ST Array 10709157 - -
          Rat Genome 230 2.0 Array 1388948_PM_at - -
          Rat Genome 230 2.0 Array 1388948_at - -
          Affymetrix HT Rat Focus Array 1388948_PM_at - -
          Affymetrix HT Rat Focus Array 1388948_at - -
          Affymetrix Rat Clariom S Assay PSR0100016714.rn.1 TC0100002165.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100016715.rn.1 TC0100002165.rn.1 RTA TC
          Rat Expression Set 230 1388948_at - -
          Rat Expression Set 230 1388948_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10709157 - -
          Rat Genome U34 Set rc_AI043768_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21286755 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline