Search
Search
| Gene Symbol: Erp29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_053961.2 | NP_446413.1 | 1 - 2 | 155 | 69 | |
| GenBank mRNA | BC091129.1 | - | 1 - 2 | 155 | 7308437 | 69 |
| FQ211664.1 | - | 1 - 2 | 195 | 69 | ||
| FQ213671.1 | - | 1 - 2 | 153 | 69 | ||
| FQ218939.1 | - | 1 - 2 | 153 | 69 | ||
| FQ220022.1 | - | 1 - 2 | 153 | 69 | ||
| FQ233694.1 | - | 1 - 2 | 153 | 69 | ||
| FQ234786.1 | - | 1 - 2 | 153 | 69 | ||
| U36482.1 | - | 1 - 2 | 183 | 69 | ||
| Y10264.1 | - | 1 - 2 | 154 | 69 | ||
Set Membership: |
|