Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00504287_g1
          See other EIF4H GE Assays ›
          Gene Symbol:
          Eif4h
          Gene Name:
          eukaryotic translation initiation factor 4H
          Gene Aliases:
          Wbscr1
          Chromosome Location:
          Chr.12: 25092926 - 25109805 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00504287_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Eif4h
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001006957.2 NP_001006958.1 6 - 7 646 88
          XM_006249134.1 XP_006249196.1 5 - 6 809 88
          GenBank mRNA BC083818.1 - 6 - 7 638 7110979 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1200000441.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Eif4h

          Panther Classification:

          Molecular Function -

          translation initiation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          translational initiation
          sexual reproduction
          developmental growth
          nucleotide binding
          translation initiation factor activity
          poly(A) RNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P833212 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 163309 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 775348 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 244273 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563025 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 289089 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 233878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45823 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 851256 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721177 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 54660 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P833212 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P833212 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P833212 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P833212 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 16849 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68587 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10757790 - -
          Affymetrix Rat Gene 1.0 ST Array 10757793 - -
          Affymetrix Rat Gene 1.0 ST Array 10757792 - -
          Affymetrix Rat Gene 1.0 ST Array 10757791 - -
          Affymetrix Rat Gene 1.0 ST Array 10757787 - -
          Affymetrix Rat Gene 1.0 ST Array 10757788 - -
          Affymetrix Rat Gene 1.0 ST Array 10757794 - -
          Affymetrix Rat Gene 1.0 ST Array 10757789 - -
          Affymetrix Rat Gene 1.1 ST Array 10757786 - -
          Rat Genome 230 2.0 Array 1367531_at - -
          Rat Genome 230 2.0 Array 1367531_PM_at - -
          Rat Genome 230 2.0 Array 1397675_at - -
          Rat Genome 230 2.0 Array 1397675_PM_at - -
          Affymetrix HT Rat Focus Array 1397675_at - -
          Affymetrix HT Rat Focus Array 1367531_at - -
          Affymetrix HT Rat Focus Array 1397675_PM_at - -
          Affymetrix HT Rat Focus Array 1367531_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1200003955.rn.1 TC1200000441.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003956.rn.1 TC1200000441.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1200003957.rn.1 TC1200000441.rn.1 RTA TC
          Rat Expression Set 230 1367531_PM_at - -
          Rat Expression Set 230 1367531_at - -
          Rat Expression Set 230 1367531_PM_at - -
          Rat Expression Set 230 1367531_at - -
          Rat Expression Set 230 1397675_PM_at - -
          Rat Expression Set 230 1397675_at - -
          Affymetrix Rat Gene 1.0 ST Array 10757786 - -
          Rat Genome U34 Set rc_AI180108_at - -
          Rat Genome U34 Set rc_AA875327_g_at - -
          Rat Genome U34 Set rc_AA875327_at - -
          Rat Genome U34 Set rc_AI059691_f_at - -
          Rat Genome U34 Set rc_AI180108_at - -
          Rat Genome U34 Set rc_AA875327_at - -
          Rat Genome U34 Set rc_AA875327_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21743932 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1373095 - -
          RatRef-12 Expression BeadChip ILMN_49914 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-8ltcg:80/100.66.77.141:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline