Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00508100_m1
          See other TMEM176B GE Assays ›
          Gene Symbol:
          Tmem176b
          Gene Name:
          transmembrane protein 176B
          Gene Aliases:
          Lr8, Torid
          Chromosome Location:
          Chr.4: 78450724 - 78458537 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00508100_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tmem176b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001270589.1 NP_001257518.1 2 - 3 828 68
          NM_001270590.1 NP_001257519.1 2 - 3 513 68
          NM_001270591.1 NP_001257520.1 3 - 4 291 68
          NM_001270592.1 NP_001257521.1 3 - 4 288 68
          NM_001270593.1 NP_001257522.1 2 - 3 225 68
          NM_001270594.1 NP_001257523.1 2 - 3 222 68
          NM_134390.3 NP_599217.1 2 - 3 831 68
          XM_006236455.2 XP_006236517.1 2 - 3 214 68
          GenBank mRNA AF370882.1 - 1 - 2 184 68
          BC059116.1 - 2 - 3 330 6920949 68
          FM070640.1 - 2 - 3 212 68
          FM086181.1 - 3 - 4 275 68
          FN804490.1 - 3 - 4 274 68
          FQ209634.1 - 2 - 3 219 68
          FQ212987.1 - 2 - 3 220 68
          FQ219149.1 - 3 - 4 275 68
          FQ219532.1 - 3 - 4 275 68
          FQ219576.1 - 2 - 3 219 68
          FQ220000.1 - 2 - 3 404 68
          FQ233370.1 - 3 - 4 285 68
          FQ235102.1 - 2 - 3 237 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0400002848.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tmem176b

          Gene Ontology Categories:

          Function(s)

          cell differentiation
          negative regulation of dendritic cell differentiation

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P139869 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 240359 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 467196 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 42852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271242 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 970532 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 44419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139869 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P139869 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P139869 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P139869 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 19360 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10862421 - -
          Affymetrix Rat Gene 1.0 ST Array 10862420 - -
          Affymetrix Rat Gene 1.0 ST Array 10862418 - -
          Affymetrix Rat Gene 1.0 ST Array 10862417 - -
          Affymetrix Rat Gene 1.0 ST Array 10862416 - -
          Affymetrix Rat Gene 1.0 ST Array 10862422 - -
          Affymetrix Rat Gene 1.0 ST Array 10862423 - -
          Affymetrix Rat Gene 1.0 ST Array 10862419 - -
          Affymetrix Rat Gene 1.1 ST Array 10862415 - -
          Rat Genome 230 2.0 Array 1368840_at - -
          Rat Genome 230 2.0 Array 1368840_PM_at - -
          Affymetrix HT Rat Focus Array 1368840_at - -
          Affymetrix HT Rat Focus Array 1368840_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0400020293.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020297.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020301.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020290.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020291.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020296.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020299.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020298.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020294.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020304.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020302.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020289.rn.1 TC0400002848.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0400020303.rn.1 TC0400002848.rn.1 RTA TC
          Rat Expression Set 230 1368840_at - -
          Rat Expression Set 230 1368840_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10862415 - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21518343 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1355909 - -
          RatRef-12 Expression BeadChip ILMN_66840 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline