Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00515007_m1
          See other GNB1 GE Assays ›
          Gene Symbol:
          Gnb1
          Gene Name:
          guanine nucleotide binding protein (G protein), beta polypeptide 1
          Gene Aliases:
          -
          Chromosome Location:
          Chr.5: 172913898 - 172981403 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00515007_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gnb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_030987.2 NP_112249.2 9 - 10 846 66
          XM_006239517.2 XP_006239579.1 9 - 10 998 66
          XM_006239518.2 XP_006239580.1 9 - 10 998 66
          XM_006239519.2 XP_006239581.1 9 - 10 998 66
          XM_006239520.2 XP_006239582.1 9 - 10 998 66
          XM_008764335.1 XP_008762557.1 8 - 9 881 66
          GenBank mRNA AY552805.1 - 7 - 8 698 66
          BC078809.1 - 9 - 10 846 7123318 66
          DQ120483.1 - 4 - 5 451 66
          DQ120484.1 - 4 - 5 451 66
          U34958.1 - 7 - 8 698 66
          U88324.1 - 7 - 8 735 66

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gnb1

          Panther Classification:

          Molecular Function -

          heterotrimeric G-protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          phospholipase C-activating G-protein coupled receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          metabolic process
          cell proliferation
          cardiac muscle cell apoptotic process
          sensory perception of taste
          retina development in camera-type eye
          cellular response to hypoxia
          G-protein coupled receptor binding
          GTPase activity
          signal transducer activity
          protein binding
          spectrin binding
          type 1 angiotensin receptor binding
          protein complex binding
          GTPase binding

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P384857 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 361209 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644840 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 956061 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298608 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312575 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 704367 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 946253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 82618 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167768 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 820778 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 756687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 330925 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 826533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313195 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 345169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 795136 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 455772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159316 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298237 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532650 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144474 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 46941 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 570660 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 926034 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 81631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167087 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45500 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 41655 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 783271 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 462215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 134507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 669729 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 271705 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 882143 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 91213 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 71290 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163457 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 173930 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 177258 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166155 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P384857 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P384857 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P384857 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468365 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P468372 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P927153 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P13986 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P802937 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700772122 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700515072 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24548 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90700 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 62088 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64433 - -
          Agilent Rat cDNA Microarray (G4105A) 700772122 - -
          Agilent Rat cDNA Microarray (G4105A) 700515072 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10874650 - -
          Affymetrix Rat Gene 1.0 ST Array 10874646 - -
          Affymetrix Rat Gene 1.0 ST Array 10874649 - -
          Affymetrix Rat Gene 1.0 ST Array 10874645 - -
          Affymetrix Rat Gene 1.0 ST Array 10874643 - -
          Affymetrix Rat Gene 1.0 ST Array 10874652 - -
          Affymetrix Rat Gene 1.0 ST Array 10874648 - -
          Affymetrix Rat Gene 1.0 ST Array 10788438 - -
          Affymetrix Rat Gene 1.0 ST Array 10874651 - -
          Affymetrix Rat Gene 1.0 ST Array 10874644 - -
          Affymetrix Rat Gene 1.0 ST Array 10874647 - -
          Affymetrix Rat Gene 1.0 ST Array 10874653 - -
          Affymetrix Rat Gene 1.0 ST Array 10874642 - -
          Affymetrix Rat Gene 1.1 ST Array 10788437 - -
          Affymetrix Rat Gene 1.1 ST Array 10874641 - -
          Rat Genome 230 2.0 Array 1367732_at - -
          Rat Genome 230 2.0 Array 1367731_PM_at - -
          Rat Genome 230 2.0 Array 1375705_at - -
          Rat Genome 230 2.0 Array 1367731_at - -
          Rat Genome 230 2.0 Array 1375705_PM_at - -
          Rat Genome 230 2.0 Array 1367732_PM_at - -
          Affymetrix HT Rat Focus Array 1367732_PM_at - -
          Affymetrix HT Rat Focus Array 1367732_at - -
          Affymetrix HT Rat Focus Array 1367731_at - -
          Affymetrix HT Rat Focus Array 1367731_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0500015657.rn.1 TC0500002063.rn.1 RTA TC
          Rat Expression Set 230 1367732_at - -
          Rat Expression Set 230 1367731_PM_at - -
          Rat Expression Set 230 1367732_PM_at - -
          Rat Expression Set 230 1367731_at - -
          Affymetrix Rat Gene 1.0 ST Array 10874641 - -
          Affymetrix Rat Gene 1.0 ST Array 10788437 - -
          Rat Genome U34 Set U88324_g_at - -
          Rat Genome U34 Set U88324_at - -
          Rat Genome U34 Set AF022083_s_at - -
          Rat Genome U34 Set rc_AI227660_s_at - -
          Rat Genome U34 Set rc_AI103622_at - -
          Rat Genome U34 Set rc_AI230404_s_at - -
          Rat Genome U34 Set U88324_at - -
          Rat Genome U34 Set rc_AI227660_s_at - -
          Rat Genome U34 Set AF022083_s_at - -
          Rat Genome U34 Set rc_AI230404_s_at - -
          Rat Genome U34 Set U88324_g_at - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1375069 - -
          RatRef-12 Expression BeadChip ILMN_49575 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline