Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00518189_m1
          See other PPP2R2D GE Assays ›
          Gene Symbol:
          Ppp2r2d
          Gene Name:
          protein phosphatase 2, regulatory subunit B, delta
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 211205903 - 211239889 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00518189_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ppp2r2d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_144746.2 NP_653347.2 2 - 3 200 98
          GenBank mRNA AF180350.1 - 2 - 3 204 98
          BC081720.1 - 2 - 3 200 7112646 98

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100002809.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ppp2r2d

          Panther Classification:

          Molecular Function -

          protein phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic nuclear division
          signal transduction
          exit from mitosis
          dephosphorylation
          regulation of catalytic activity
          cell division
          phosphatidate phosphatase activity
          protein phosphatase type 2A regulator activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P112724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 203080 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757903 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 136875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 273349 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 170108 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 523477 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371338 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 778445 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 635854 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 955009 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 690266 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705968 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 206280 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949546 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 168853 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 68023 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 828923 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 967253 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543914 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405577 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413181 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 880233 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166574 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 23076 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 25434 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 12179 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112724 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P112724 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P112724 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P249531 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701624832 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701520563 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700630591 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701623170 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 55214 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 13952 - -
          Agilent Rat cDNA Microarray (G4105A) 700630591 - -
          Agilent Rat cDNA Microarray (G4105A) 701624832 - -
          Agilent Rat cDNA Microarray (G4105A) 701520563 - -
          Agilent Rat cDNA Microarray (G4105A) 701623170 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10711904 - -
          Affymetrix Rat Gene 1.0 ST Array 10711898 - -
          Affymetrix Rat Gene 1.0 ST Array 10711905 - -
          Affymetrix Rat Gene 1.0 ST Array 10711900 - -
          Affymetrix Rat Gene 1.0 ST Array 10711896 - -
          Affymetrix Rat Gene 1.0 ST Array 10711895 - -
          Affymetrix Rat Gene 1.0 ST Array 10711902 - -
          Affymetrix Rat Gene 1.0 ST Array 10711901 - -
          Affymetrix Rat Gene 1.0 ST Array 10711903 - -
          Affymetrix Rat Gene 1.0 ST Array 10711897 - -
          Affymetrix Rat Gene 1.0 ST Array 10711899 - -
          Affymetrix Rat Gene 1.1 ST Array 10711894 - -
          Rat Genome 230 2.0 Array 1388107_PM_at - -
          Rat Genome 230 2.0 Array 1388107_at - -
          Affymetrix HT Rat Focus Array 1388107_PM_at - -
          Affymetrix HT Rat Focus Array 1388107_at - -
          Affymetrix Rat Clariom S Assay PSR0100022106.rn.1 TC0100002809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022103.rn.1 TC0100002809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022108.rn.1 TC0100002809.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0100022107.rn.1 TC0100002809.rn.1 RTA TC
          Rat Expression Set 230 1388107_PM_at - -
          Rat Expression Set 230 1388107_at - -
          Affymetrix Rat Gene 1.0 ST Array 10711894 - -
          Rat Genome U34 Set rc_AI029941_at - -
          Rat Genome U34 Set rc_AI177774_at - -
          Rat Genome U34 Set rc_AI234973_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21089421 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1371291 - -
          RatRef-12 Expression BeadChip ILMN_52415 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline