Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00524612_m1
          See other GABRP GE Assays ›
          Gene Symbol:
          Gabrp
          Gene Name:
          gamma-aminobutyric acid (GABA-A) receptor, pi
          Gene Aliases:
          -
          Chromosome Location:
          Chr.10: 18484257 - 18510606 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00524612_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Gabrp
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_031029.1 NP_112291.1 8 - 9 1063 94
          XM_008767594.1 XP_008765816.1 9 - 10 1649 94
          XM_008767595.1 XP_008765817.1 9 - 10 1655 94
          GenBank mRNA FQ222085.1 - 9 - 10 1230 94
          U95368.1 - 8 - 9 1063 94

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000002516.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Gabrp

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          signal transduction
          ion transmembrane transport
          chloride transmembrane transport
          GABA-A receptor activity
          extracellular ligand-gated ion channel activity
          chloride channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P326917 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 465682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 380337 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 20118 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 772622 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 721092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 744842 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 861953 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 414282 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 402610 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 122559 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 103720 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842414 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 167443 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 125817 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P326917 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P326917 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P326917 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P326917 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 42812 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 82929 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10741952 - -
          Affymetrix Rat Gene 1.0 ST Array 10741951 - -
          Affymetrix Rat Gene 1.0 ST Array 10741953 - -
          Affymetrix Rat Gene 1.0 ST Array 10741948 - -
          Affymetrix Rat Gene 1.0 ST Array 10741955 - -
          Affymetrix Rat Gene 1.0 ST Array 10741950 - -
          Affymetrix Rat Gene 1.0 ST Array 10741949 - -
          Affymetrix Rat Gene 1.1 ST Array 10741947 - -
          Rat Genome 230 2.0 Array 1387600_at - -
          Rat Genome 230 2.0 Array 1387600_PM_at - -
          Affymetrix HT Rat Focus Array 1387600_PM_at - -
          Affymetrix HT Rat Focus Array 1387600_at - -
          Affymetrix Rat Clariom S Assay PSR1000022802.rn.1 TC1000002516.rn.1 RTA TC
          Rat Expression Set 230 1387600_PM_at - -
          Rat Expression Set 230 1387600_at - -
          Affymetrix Rat Gene 1.0 ST Array 10741947 - -
          Rat Genome U34 Set U95368_at - -
          Rat Genome U34 Set U95368_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22331341 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1368693 - -
          RatRef-12 Expression BeadChip ILMN_67368 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline