Search
Search
| Gene Symbol: Dctn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004239.1 | NP_001004239.1 | 1 - 2 | 144 | 74 | |
| XM_006241448.1 | XP_006241510.1 | 1 - 2 | 226 | 74 | ||
| XM_008765392.1 | XP_008763614.1 | 1 - 2 | 228 | 74 | ||
| GenBank mRNA | BC079042.1 | - | 1 - 2 | 144 | 7114929 | 74 |
Set Membership: |
|