Search
Search
| Gene Symbol: Afp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012493.2 | NP_036625.2 | 3 - 4 | 296 | 81 | |
| GenBank mRNA | BC097344.1 | - | 3 - 4 | 279 | 7459964 | 81 |
| DY309525.1 | - | 3 - 4 | 296 | 81 | ||
| X02361.1 | - | 3 - 4 | 300 | 81 | ||
Set Membership: |
|