Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00560747_m1
          See other AR GE Assays ›
          Gene Symbol:
          Ar
          Gene Name:
          androgen receptor
          Gene Aliases:
          Andr, Tfm
          Chromosome Location:
          Chr.X: 67656253 - 67828998 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00560747_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information
          • Citations (1)

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ar
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012502.1 NP_036634.1 4 - 5 3115 75
          GenBank mRNA M20133.1 - 4 - 5 3115 75
          M23264.1 - 4 - 5 2154 75

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0X00000570.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Recommended Assay Rattus norvegicus Rat Androgen Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ar

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          regulation of systemic arterial blood pressure
          epithelial cell morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          spermatogenesis
          single fertilization
          positive regulation of cell proliferation
          male gonad development
          fertilization
          cellular process
          regulation of gene expression
          male somatic sex determination
          androgen receptor signaling pathway
          intracellular receptor signaling pathway
          response to insulin
          positive regulation of intracellular estrogen receptor signaling pathway
          Leydig cell differentiation
          multicellular organism growth
          positive regulation of phosphorylation
          steroid hormone mediated signaling pathway
          positive regulation of MAPK cascade
          positive regulation of insulin-like growth factor receptor signaling pathway
          negative regulation of integrin biosynthetic process
          positive regulation of integrin biosynthetic process
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of transcription from RNA polymerase III promoter
          male sex differentiation
          reproductive structure development
          regulation of developmental growth
          organ formation
          male genitalia morphogenesis
          negative regulation of epithelial cell proliferation
          regulation of catalytic activity
          positive regulation of NF-kappaB transcription factor activity
          protein oligomerization
          activation of prostate induction by androgen receptor signaling pathway
          morphogenesis of an epithelial fold
          lateral sprouting involved in mammary gland duct morphogenesis
          regulation of prostatic bud formation
          prostate gland growth
          prostate gland epithelium morphogenesis
          epithelial cell differentiation involved in prostate gland development
          tertiary branching involved in mammary gland duct morphogenesis
          mammary gland alveolus development
          reproductive system development
          cellular response to steroid hormone stimulus
          seminiferous tubule development
          regulation of establishment of protein localization to plasma membrane
          negative regulation of extrinsic apoptotic signaling pathway
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          androgen receptor activity
          receptor binding
          androgen binding
          protein binding
          beta-catenin binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein domain specific binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          androgen receptor binding
          POU domain binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P037631 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 416724 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 15942 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291927 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315732 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 790467 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 97098 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125363 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401225 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 490970 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 272852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587217 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 566773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 524055 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644090 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 458508 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 230169 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 442422 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 216214 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587102 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331843 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 153908 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 553354 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 653905 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 814451 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 204287 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 799174 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 971376 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 487928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 148977 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 802875 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 9730 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 450841 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 466606 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 515806 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393813 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 358431 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 354743 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 328531 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 119605 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688887 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 655716 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 315155 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 175679 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 167528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 809620 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 351986 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 443187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 65625 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 107111 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 381816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 210092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 637113 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 77372 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104222 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 294948 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449671 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 761913 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943772 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 856989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 757004 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724326 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 331523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 376599 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 445703 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934672 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 921407 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 71619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 212115 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648292 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 564573 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143871 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 288027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 160669 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 471636 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 52350 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249549 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 965684 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 932305 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 60537 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 660847 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 587227 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 85912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 262364 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393971 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 512598 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 877533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279528 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 95755 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 558619 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 382685 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 760788 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 751989 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 684492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 270438 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 231232 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810092 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 628719 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 964588 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 785340 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 705210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 689858 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 475502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 468024 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 847360 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312939 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 842251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521785 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149345 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 741890 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 534021 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 45796 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 944308 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 969297 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 810956 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 101313 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 409674 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 567260 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 201086 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 427687 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 673782 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 844877 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688130 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 729134 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 688352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 478604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 737461 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781556 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 191187 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 323088 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 104628 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 840663 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121407 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 55419 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 153852 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 114556 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 117851 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 95358 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 38092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 149303 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 34910 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 13440 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 39004 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 137847 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 70656 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 180405 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 14120 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 142299 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 151188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 174097 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83322 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 80879 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 166201 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118866 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037631 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P037631 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P037631 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P546901 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 10764 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 78337 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32992 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46985 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75280 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64744 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 24302 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 8670 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 68499 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 15538 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 758 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 66938 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 38617 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10934121 - -
          Affymetrix Rat Gene 1.0 ST Array 10934123 - -
          Affymetrix Rat Gene 1.0 ST Array 10934126 - -
          Affymetrix Rat Gene 1.0 ST Array 10934119 - -
          Affymetrix Rat Gene 1.0 ST Array 10934125 - -
          Affymetrix Rat Gene 1.0 ST Array 10934122 - -
          Affymetrix Rat Gene 1.0 ST Array 10934120 - -
          Affymetrix Rat Gene 1.0 ST Array 10934124 - -
          Affymetrix Rat Gene 1.0 ST Array 10934127 - -
          Affymetrix Rat Gene 1.1 ST Array 10934118 - -
          Rat Genome 230 2.0 Array 1369159_at - -
          Rat Genome 230 2.0 Array 1369159_PM_at - -
          Affymetrix HT Rat Focus Array 1369159_at - -
          Affymetrix HT Rat Focus Array 1369159_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0X00003514.rn.1 TC0X00000570.rn.1 RTA TC
          Rat Expression Set 230 1369159_PM_at - -
          Rat Expression Set 230 1369159_at - -
          Affymetrix Rat Gene 1.0 ST Array 10934118 - -
          Rat Genome U34 Set M23264_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21272180 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1353640 - -
          RatRef-12 Expression BeadChip ILMN_68782 - -

          Back To Top
           

          Citations (1)

          Androgen-dependent mechanisms of Wolffian duct development and their perturbation by flutamide.
          Authors
          Welsh M, Saunders PT, Marchetti NI, Sharpe RM
          Journal
          Endocrinology - Page No 4820 (Sun Oct 01 00:00:00 EDT 2006)
          Volume (Year)
          147 (2006)
          Product Usage
          -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline