Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00560813_m1
          See other ATP1A3 GE Assays ›
          Gene Symbol:
          Atp1a3
          Gene Name:
          ATPase, Na+/K+ transporting, alpha 3 polypeptide
          Gene Aliases:
          Atpa1a3
          Chromosome Location:
          Chr.1: 81852425 - 81881549 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00560813_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp1a3
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012506.1 NP_036638.1 4 - 5 504 63
          XM_008758888.1 XP_008757110.1 4 - 5 418 63
          GenBank mRNA FQ212331.1 - 3 - 4 341 63
          M14513.1 - 4 - 5 504 63
          X05883.1 - 4 - 5 501 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0100004979.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp1a3

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          potassium ion transport
          sodium ion transport
          cellular sodium ion homeostasis
          memory
          metabolic process
          adult locomotory behavior
          visual learning
          potassium ion import
          ATP hydrolysis coupled proton transport
          cellular potassium ion homeostasis
          ionotropic glutamate receptor signaling pathway
          sodium ion export from cell
          response to drug
          sodium:potassium-exchanging ATPase activity
          protein binding
          ATP binding
          cation-transporting ATPase activity
          D1 dopamine receptor binding
          metal ion binding
          chaperone binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P400177 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 692013 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 298205 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 476677 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227479 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 700653 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 149459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 275682 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 664816 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 513928 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 405016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 211383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 862344 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 953142 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961698 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531934 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 54485 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 616125 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227973 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 781459 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 952002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 743707 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 302436 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258342 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 68188 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 32069 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 121858 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 35001 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163761 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400177 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P400177 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P400177 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P400177 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 75814 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 11846 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10719743 - -
          Affymetrix Rat Gene 1.0 ST Array 10719741 - -
          Affymetrix Rat Gene 1.0 ST Array 10719750 - -
          Affymetrix Rat Gene 1.0 ST Array 10719737 - -
          Affymetrix Rat Gene 1.0 ST Array 10719730 - -
          Affymetrix Rat Gene 1.0 ST Array 10719749 - -
          Affymetrix Rat Gene 1.0 ST Array 10719738 - -
          Affymetrix Rat Gene 1.0 ST Array 10719747 - -
          Affymetrix Rat Gene 1.0 ST Array 10719739 - -
          Affymetrix Rat Gene 1.0 ST Array 10719742 - -
          Affymetrix Rat Gene 1.0 ST Array 10719740 - -
          Affymetrix Rat Gene 1.0 ST Array 10719748 - -
          Affymetrix Rat Gene 1.0 ST Array 10719734 - -
          Affymetrix Rat Gene 1.0 ST Array 10719733 - -
          Affymetrix Rat Gene 1.0 ST Array 10719746 - -
          Affymetrix Rat Gene 1.0 ST Array 10719752 - -
          Affymetrix Rat Gene 1.0 ST Array 10719729 - -
          Affymetrix Rat Gene 1.0 ST Array 10719751 - -
          Affymetrix Rat Gene 1.0 ST Array 10719732 - -
          Affymetrix Rat Gene 1.0 ST Array 10719745 - -
          Affymetrix Rat Gene 1.0 ST Array 10719753 - -
          Affymetrix Rat Gene 1.0 ST Array 10719735 - -
          Affymetrix Rat Gene 1.0 ST Array 10719744 - -
          Affymetrix Rat Gene 1.0 ST Array 10719736 - -
          Affymetrix Rat Gene 1.1 ST Array 10719728 - -
          Rat Genome 230 2.0 Array 1368701_at - -
          Rat Genome 230 2.0 Array 1368701_PM_at - -
          Affymetrix HT Rat Focus Array 1368701_at - -
          Affymetrix HT Rat Focus Array 1368701_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0100039487.rn.1 TC0100004979.rn.1 RTA TC
          Rat Expression Set 230 1368701_PM_at - -
          Rat Expression Set 230 1368701_at - -
          Affymetrix Rat Gene 1.0 ST Array 10719728 - -
          Rat Genome U34 Set M28648_s_at - -
          Rat Genome U34 Set D00189_at - -
          Rat Genome U34 Set M28648_s_at - -
          Rat Genome U34 Set M28648_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21945469 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1361151 - -
          RatRef-12 Expression BeadChip ILMN_47976 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline