Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00560862_m1
          See other ATP7B GE Assays ›
          Gene Symbol:
          Atp7b
          Gene Name:
          ATPase, Cu++ transporting, beta polypeptide
          Gene Aliases:
          Hts, PINA, Wd
          Chromosome Location:
          Chr.16: 74865516 - 74945956 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00560862_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Atp7b
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012511.2 NP_036643.2 7 - 8 2099 88
          XM_006253346.2 XP_006253408.1 7 - 8 2775 88
          GenBank mRNA AB017790.1 - 7 - 8 2099 88
          AB017791.1 - 7 - 8 2099 88
          AB017792.1 - 7 - 8 2099 88
          AB017793.1 - 7 - 8 2099 88
          U08344.1 - 7 - 8 2115 88

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1600000886.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Transporter Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Atp7b

          Panther Classification:

          Molecular Function -

          primary active transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          copper ion transport
          cellular copper ion homeostasis
          cellular zinc ion homeostasis
          lactation
          circadian rhythm
          metabolic process
          response to zinc ion
          copper ion import
          intracellular copper ion transport
          metal ion transport
          copper ion transmembrane transport
          response to copper ion
          sequestering of calcium ion
          response to cAMP
          copper ion export
          cation transmembrane transport
          copper-exporting ATPase activity
          copper ion transmembrane transporter activity
          copper ion binding
          ATP binding
          zinc ion binding
          cation-transporting ATPase activity
          copper-transporting ATPase activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017640 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P017645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 159148 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 581803 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608262 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 521105 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 397129 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 291294 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147892 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 312094 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 366002 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544756 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 91006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590027 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 37557 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 449251 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 371006 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 759455 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128375 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 531081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 454492 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 545361 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 156735 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 527946 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 678265 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 128645 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 125060 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 224443 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 590929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 624569 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 565388 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 910215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 343826 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 53799 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 724714 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 884502 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 593408 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 304844 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 577929 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 256507 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 648268 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 360081 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 575765 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 359277 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 143808 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 868245 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 787909 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 697471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 663859 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 784795 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 22351 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 134462 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 96494 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 46601 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 83774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 124252 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 18391 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 118092 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 164456 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 56036 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017645 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017640 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P017645 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017645 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P017640 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P306339 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 40799 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 83055 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 26138 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 72637 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 46481 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10789220 - -
          Affymetrix Rat Gene 1.0 ST Array 10789217 - -
          Affymetrix Rat Gene 1.0 ST Array 10789201 - -
          Affymetrix Rat Gene 1.0 ST Array 10789208 - -
          Affymetrix Rat Gene 1.0 ST Array 10789211 - -
          Affymetrix Rat Gene 1.0 ST Array 10789204 - -
          Affymetrix Rat Gene 1.0 ST Array 10789216 - -
          Affymetrix Rat Gene 1.0 ST Array 10789212 - -
          Affymetrix Rat Gene 1.0 ST Array 10789200 - -
          Affymetrix Rat Gene 1.0 ST Array 10789202 - -
          Affymetrix Rat Gene 1.0 ST Array 10789207 - -
          Affymetrix Rat Gene 1.0 ST Array 10789213 - -
          Affymetrix Rat Gene 1.0 ST Array 10789206 - -
          Affymetrix Rat Gene 1.0 ST Array 10789214 - -
          Affymetrix Rat Gene 1.0 ST Array 10789219 - -
          Affymetrix Rat Gene 1.0 ST Array 10789221 - -
          Affymetrix Rat Gene 1.0 ST Array 10789223 - -
          Affymetrix Rat Gene 1.0 ST Array 10789209 - -
          Affymetrix Rat Gene 1.0 ST Array 10789218 - -
          Affymetrix Rat Gene 1.0 ST Array 10789203 - -
          Affymetrix Rat Gene 1.0 ST Array 10789205 - -
          Affymetrix Rat Gene 1.0 ST Array 10789215 - -
          Affymetrix Rat Gene 1.0 ST Array 10789222 - -
          Affymetrix Rat Gene 1.0 ST Array 10789210 - -
          Affymetrix Rat Gene 1.1 ST Array 10789199 - -
          Rat Genome 230 2.0 Array 1370324_PM_at - -
          Rat Genome 230 2.0 Array 1369123_PM_a_at - -
          Rat Genome 230 2.0 Array 1370324_at - -
          Rat Genome 230 2.0 Array 1369123_a_at - -
          Affymetrix HT Rat Focus Array 1369123_PM_a_at - -
          Affymetrix HT Rat Focus Array 1370324_at - -
          Affymetrix HT Rat Focus Array 1369123_a_at - -
          Affymetrix HT Rat Focus Array 1370324_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1600006877.rn.1 TC1600000886.rn.1 RTA TC
          Rat Expression Set 230 1369123_a_at - -
          Rat Expression Set 230 1370324_PM_at - -
          Rat Expression Set 230 1369123_PM_a_at - -
          Rat Expression Set 230 1370324_at - -
          Affymetrix Rat Gene 1.0 ST Array 10789199 - -
          Rat Genome U34 Set U08344_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22273521 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1357509 - -
          RatRef-12 Expression BeadChip ILMN_49164 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline