Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00560892_m1
          See other TSPO GE Assays ›
          Gene Symbol:
          Tspo
          Gene Name:
          translocator protein
          Gene Aliases:
          Bzrp, MBR, PTBZR02, Ptbzr, RATPTBZR02
          Chromosome Location:
          Chr.7: 124460358 - 124470610 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00560892_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Tspo
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012515.2 NP_036647.1 3 - 4 411 117
          GenBank mRNA BQ194961.1 - 1 - 2 117
          FQ228784.1 - 3 - 4 413 117
          J05122.1 - 2 - 3 354 117

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0700001553.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Transporter 3' Most Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Tspo

          Gene Ontology Categories:

          Function(s) Process(es)

          steroid biosynthetic process
          ion transport
          chloride transport
          lipid transport
          chemical synaptic transmission
          aging
          glial cell migration
          response to manganese ion
          response to vitamin B1
          positive regulation of necrotic cell death
          peripheral nervous system axon regeneration
          adrenal gland development
          response to progesterone
          negative regulation of tumor necrosis factor production
          response to testosterone
          response to drug
          positive regulation of apoptotic process
          negative regulation of nitric oxide biosynthetic process
          response to pain
          behavioral response to pain
          response to axon injury
          regulation of steroid biosynthetic process
          positive regulation of mitochondrial depolarization
          positive regulation of calcium ion transport
          contact inhibition
          positive regulation of glial cell proliferation
          negative regulation of glial cell proliferation
          cellular response to lipopolysaccharide
          cellular response to zinc ion
          cellular hypotonic response
          positive regulation of reactive oxygen species metabolic process
          androgen binding
          protein binding
          benzodiazepine receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_42_P544601 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 627791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 876008 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P544601 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_42_P544601 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_42_P544601 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P514939 - -
          Agilent Rat cDNA Microarray (G4105A) 700544254 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10898195 - -
          Affymetrix Rat Gene 1.0 ST Array 10898194 - -
          Affymetrix Rat Gene 1.0 ST Array 10898193 - -
          Affymetrix Rat Gene 1.1 ST Array 10898192 - -
          Rat Genome 230 2.0 Array 1370249_at - -
          Rat Genome 230 2.0 Array 1370249_PM_at - -
          Affymetrix HT Rat Focus Array 1370249_PM_at - -
          Affymetrix HT Rat Focus Array 1370249_at - -
          Affymetrix Rat Clariom S Assay PSR0700011816.rn.1 TC0700001553.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0700011817.rn.1 TC0700001553.rn.1 RTA TC
          Rat Expression Set 230 1370249_PM_at - -
          Rat Expression Set 230 1370249_at - -
          Affymetrix Rat Gene 1.0 ST Array 10898192 - -
          Rat Genome U34 Set J05122_at - -
          Rat Genome U34 Set J05122_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21668081 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1349787 - -
          RatRef-12 Expression BeadChip ILMN_55731 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline