Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561021_m1
          See other CHRNB1 GE Assays ›
          Gene Symbol:
          Chrnb1
          Gene Name:
          cholinergic receptor, nicotinic, beta 1 (muscle)
          Gene Aliases:
          Acrb, RNACRB1
          Chromosome Location:
          Chr.10: 56390671 - 56403226 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561021_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Chrnb1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012528.1 NP_036660.1 8 - 9 1113 59
          XM_008767770.1 XP_008765992.1 8 - 9 1160 59
          GenBank mRNA X74833.1 - 8 - 9 1113 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000003095.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Chrnb1

          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          postsynaptic membrane organization
          skeletal muscle contraction
          cation transport
          muscle contraction
          signal transduction
          synaptic transmission, cholinergic
          neuromuscular synaptic transmission
          ion transmembrane transport
          behavioral response to nicotine
          regulation of membrane potential
          muscle fiber development
          neurological system process
          transmembrane transport
          cation transmembrane transport
          acetylcholine-activated cation-selective channel activity
          channel activity
          ligand-gated ion channel activity
          acetylcholine receptor activity
          acetylcholine binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P082893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 609343 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 683864 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 19082 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 891033 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 685791 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 29555 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 335259 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 344194 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 165412 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082893 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P082893 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P082893 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P693011 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 90415 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 88704 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10744219 - -
          Affymetrix Rat Gene 1.0 ST Array 10744225 - -
          Affymetrix Rat Gene 1.0 ST Array 10744218 - -
          Affymetrix Rat Gene 1.0 ST Array 10744226 - -
          Affymetrix Rat Gene 1.0 ST Array 10744221 - -
          Affymetrix Rat Gene 1.0 ST Array 10744216 - -
          Affymetrix Rat Gene 1.0 ST Array 10744217 - -
          Affymetrix Rat Gene 1.0 ST Array 10744222 - -
          Affymetrix Rat Gene 1.0 ST Array 10744220 - -
          Affymetrix Rat Gene 1.0 ST Array 10744223 - -
          Affymetrix Rat Gene 1.0 ST Array 10744224 - -
          Affymetrix Rat Gene 1.1 ST Array 10744215 - -
          Rat Genome 230 2.0 Array 1369091_at - -
          Rat Genome 230 2.0 Array 1379870_PM_at - -
          Rat Genome 230 2.0 Array 1369091_PM_at - -
          Rat Genome 230 2.0 Array 1379870_at - -
          Affymetrix HT Rat Focus Array 1379870_PM_at - -
          Affymetrix HT Rat Focus Array 1369091_at - -
          Affymetrix HT Rat Focus Array 1379870_at - -
          Affymetrix HT Rat Focus Array 1369091_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000027617.rn.1 TC1000003095.rn.1 RTA TC
          Rat Expression Set 230 1369091_at - -
          Rat Expression Set 230 1369091_PM_at - -
          Rat Expression Set 230 1379870_at - -
          Rat Expression Set 230 1379870_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10744215 - -
          Rat Genome U34 Set X74833cds_at - -
          Rat Genome U34 Set X74833cds_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21743334 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1372351 - -
          RatRef-12 Expression BeadChip ILMN_48316 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline