Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561025_g1
          See other CKB GE Assays ›
          Gene Symbol:
          Ckb
          Gene Name:
          creatine kinase, brain
          Gene Aliases:
          Ckbb, Ckbr
          Chromosome Location:
          Chr.6: 136142956 - 136145838 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561025_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ckb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012529.3 NP_036661.3 565 59
          GenBank mRNA BC065307.1 - 547 5598839 59
          BC070955.1 - 552 7104205 59
          BC087656.1 - 564 59
          BC099814.1 - 549 7929525 59
          BC127477.1 - 555 8367180 59
          FQ216091.1 - 2 - 3 284 59
          M14400.1 - 577 59
          M57664.1 - 3 - 4 428 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0600003219.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ckb

          Panther Classification:

          Molecular Function -

          amino acid kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          phosphocreatine metabolic process
          brain development
          phosphorylation
          substantia nigra development
          cellular chloride ion homeostasis
          creatine kinase activity
          protein binding
          ATP binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_44_P264299 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 279957 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 347110 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264299 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_44_P264299 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_44_P264299 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P264299 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700147686 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79655 - -
          Agilent Rat cDNA Microarray (G4105A) 700147686 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10892269 - -
          Affymetrix Rat Gene 1.0 ST Array 10892270 - -
          Affymetrix Rat Gene 1.0 ST Array 10892267 - -
          Affymetrix Rat Gene 1.0 ST Array 10892272 - -
          Affymetrix Rat Gene 1.0 ST Array 10892268 - -
          Affymetrix Rat Gene 1.0 ST Array 10892266 - -
          Affymetrix Rat Gene 1.0 ST Array 10892271 - -
          Affymetrix Rat Gene 1.0 ST Array 10892273 - -
          Affymetrix Rat Gene 1.1 ST Array 10892265 - -
          Rat Genome 230 2.0 Array 1367740_PM_at - -
          Rat Genome 230 2.0 Array 1367740_at - -
          Affymetrix HT Rat Focus Array 1367740_at - -
          Affymetrix HT Rat Focus Array 1367740_PM_at - -
          Affymetrix Rat Clariom S Assay PSR0600022773.rn.1 TC0600003219.rn.1 RTA TC
          Rat Expression Set 230 1367740_PM_at - -
          Rat Expression Set 230 1367740_at - -
          Affymetrix Rat Gene 1.0 ST Array 10892265 - -
          Rat Genome U34 Set M57664_g_at - -
          Rat Genome U34 Set M57664_at - -
          Rat Genome U34 Set rc_AI227961_f_at - -
          Rat Genome U34 Set M57664_at - -
          Rat Genome U34 Set M57664_g_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20709867 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1370888 - -
          RatRef-12 Expression BeadChip ILMN_58424 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline