Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561094_m1
          See other ACE GE Assays ›
          Gene Symbol:
          Ace
          Gene Name:
          angiotensin I converting enzyme
          Gene Aliases:
          Dcp1, StsRR92
          Chromosome Location:
          Chr.10: 94170766 - 94213831 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561094_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ace
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012544.1 NP_036676.1 15 - 16 2344 90
          GenBank mRNA AF201331.1 - 15 - 16 2344 90
          AF201332.1 - 15 - 16 2344 90
          AF532783.1 - 15 - 16 2288 90
          AF532784.1 - 15 - 16 2288 90
          AF539425.1 - 721 90
          BC085760.1 - 15 - 16 2344 7097136 90
          L36664.1 - 2 - 3 148 90
          U03708.1 - 15 - 16 2344 90
          U03734.1 - 15 - 16 2344 90

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004380.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Protease Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ace

          Panther Classification:

          Molecular Function -

          metalloprotease

          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          kidney development
          neutrophil mediated immunity
          regulation of systemic arterial blood pressure by renin-angiotensin
          positive regulation of systemic arterial blood pressure
          proteolysis
          peptide metabolic process
          spermatogenesis
          brain development
          female pregnancy
          aging
          regulation of blood pressure
          male gonad development
          posttranscriptional regulation of gene expression
          negative regulation of gene expression
          bradykinin catabolic process
          regulation of smooth muscle cell migration
          sensory perception of pain
          lung development
          organ regeneration
          response to nutrient levels
          negative regulation of protein binding
          positive regulation of protein binding
          response to lipopolysaccharide
          response to laminar fluid shear stress
          vasoconstriction
          hormone catabolic process
          response to drug
          eating behavior
          positive regulation of apoptotic process
          peptide catabolic process
          lung alveolus development
          beta-amyloid metabolic process
          arachidonic acid secretion
          positive regulation of inflammatory response
          positive regulation of neurogenesis
          heart contraction
          regulation of angiotensin metabolic process
          angiogenesis involved in coronary vascular morphogenesis
          positive regulation of protein tyrosine kinase activity
          cellular response to glucose stimulus
          response to dexamethasone
          cell proliferation in bone marrow
          negative regulation of calcium ion import
          response to thyroid hormone
          positive regulation of peptidyl-tyrosine autophosphorylation
          regulation of hematopoietic stem cell proliferation
          negative regulation of gap junction assembly
          positive regulation of peptidyl-cysteine S-nitrosylation
          actin binding
          endopeptidase activity
          carboxypeptidase activity
          drug binding
          peptidase activity
          metallopeptidase activity
          exopeptidase activity
          tripeptidyl-peptidase activity
          peptidyl-dipeptidase activity
          zinc ion binding
          chloride ion binding
          mitogen-activated protein kinase kinase binding
          bradykinin receptor binding
          metal ion binding
          mitogen-activated protein kinase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P010531 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P098230 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608215 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 303604 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413334 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 398856 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 943189 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 186978 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 770609 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 387902 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 373834 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 532153 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 625879 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 651447 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 365852 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 258398 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 88430 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42918 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 106388 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010531 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P098230 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P010531 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P098230 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P010531 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11457 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P513075 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700308255 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 53629 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95681 - -
          Agilent Rat cDNA Microarray (G4105A) 700308255 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10739047 - -
          Affymetrix Rat Gene 1.0 ST Array 10739037 - -
          Affymetrix Rat Gene 1.0 ST Array 10739056 - -
          Affymetrix Rat Gene 1.0 ST Array 10739038 - -
          Affymetrix Rat Gene 1.0 ST Array 10739065 - -
          Affymetrix Rat Gene 1.0 ST Array 10739045 - -
          Affymetrix Rat Gene 1.0 ST Array 10739055 - -
          Affymetrix Rat Gene 1.0 ST Array 10739052 - -
          Affymetrix Rat Gene 1.0 ST Array 10739058 - -
          Affymetrix Rat Gene 1.0 ST Array 10739042 - -
          Affymetrix Rat Gene 1.0 ST Array 10739062 - -
          Affymetrix Rat Gene 1.0 ST Array 10739064 - -
          Affymetrix Rat Gene 1.0 ST Array 10739060 - -
          Affymetrix Rat Gene 1.0 ST Array 10739046 - -
          Affymetrix Rat Gene 1.0 ST Array 10739041 - -
          Affymetrix Rat Gene 1.0 ST Array 10739051 - -
          Affymetrix Rat Gene 1.0 ST Array 10739040 - -
          Affymetrix Rat Gene 1.0 ST Array 10739053 - -
          Affymetrix Rat Gene 1.0 ST Array 10739049 - -
          Affymetrix Rat Gene 1.0 ST Array 10739044 - -
          Affymetrix Rat Gene 1.0 ST Array 10739063 - -
          Affymetrix Rat Gene 1.0 ST Array 10739059 - -
          Affymetrix Rat Gene 1.0 ST Array 10739066 - -
          Affymetrix Rat Gene 1.0 ST Array 10739061 - -
          Affymetrix Rat Gene 1.0 ST Array 10739057 - -
          Affymetrix Rat Gene 1.0 ST Array 10739043 - -
          Affymetrix Rat Gene 1.0 ST Array 10739039 - -
          Affymetrix Rat Gene 1.0 ST Array 10739036 - -
          Affymetrix Rat Gene 1.1 ST Array 10739035 - -
          Rat Genome 230 2.0 Array 1370028_at - -
          Rat Genome 230 2.0 Array 1381062_at - -
          Rat Genome 230 2.0 Array 1387791_at - -
          Rat Genome 230 2.0 Array 1370028_PM_at - -
          Rat Genome 230 2.0 Array 1381062_PM_at - -
          Rat Genome 230 2.0 Array 1387791_PM_at - -
          Affymetrix HT Rat Focus Array 1387791_PM_at - -
          Affymetrix HT Rat Focus Array 1370028_PM_at - -
          Affymetrix HT Rat Focus Array 1370028_at - -
          Affymetrix HT Rat Focus Array 1387791_at - -
          Affymetrix Rat Clariom S Assay PSR1000040832.rn.1 TC1000004380.rn.1 RTA TC
          Rat Expression Set 230 1387791_PM_at - -
          Rat Expression Set 230 1370028_at - -
          Rat Expression Set 230 1370028_PM_at - -
          Rat Expression Set 230 1387791_at - -
          Affymetrix Rat Gene 1.0 ST Array 10739035 - -
          Rat Genome U34 Set L36664_s_at - -
          Rat Genome U34 Set U03734_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 21301215 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1362259 - -
          RatRef-12 Expression BeadChip ILMN_56634 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline