Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Pipettes and Pipette Tips
    • Lab Centrifuges
    • Ultra-Low Temperature Freezers
    • Spectroscopy
    • Beakers
    • PCR Equipment and Supplies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Cell Analysis
    • Lab Equipment
    • Real-Time PCR
    • PCR
    • Chromatography
    • Cell Culture and Transfection
    • DNA and RNA Extraction and Analysis
    • Protein Biology
    • Flow Cytometry
    • Chemicals
    • See all applications and techniques
  • Services
    • Custom Services
    • Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Instrument Services
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561137_m1
          See other EDNRA GE Assays ›
          Gene Symbol:
          Ednra
          Gene Name:
          endothelin receptor type A
          Gene Aliases:
          ET-A, ET-AR, Endor, Eta, RGD1559432
          Chromosome Location:
          Chr.19: 33928190 - 33991703 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561137_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ednra
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012550.2 NP_036682.1 2 - 3 715 118
          XM_006255420.2 XP_006255482.1 2 - 3 881 118
          XM_008772471.1 XP_008770693.1 2 - 3 588 118
          GenBank mRNA DQ832324.1 - 1 - 2 466 118
          DQ839233.1 - 1 - 2 449 118
          M60786.1 - 1 - 2 466 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1900000437.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 G-protein coupled receptor 3' Most Recommended Assay Rattus norvegicus Rat Hypoxia Panel 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ednra

          Gene Ontology Categories:

          Function(s) Process(es)

          patterning of blood vessels
          response to hypoxia
          in utero embryonic development
          histamine secretion
          positive regulation of protein phosphorylation
          glomerular filtration
          positive regulation of cytosolic calcium ion concentration
          protein kinase C-activating G-protein coupled receptor signaling pathway
          Rho protein signal transduction
          heart development
          aging
          respiratory gaseous exchange
          regulation of blood pressure
          positive regulation of cell proliferation
          neural crest cell development
          response to organic cyclic compound
          artery smooth muscle contraction
          glucose transport
          sensory perception of pain
          negative regulation of cAMP biosynthetic process
          response to lipopolysaccharide
          vasoconstriction
          positive regulation of odontogenesis
          negative regulation of apoptotic process
          penile erection
          response to morphine
          fibroblast proliferation
          enteric nervous system development
          smooth muscle cell proliferation
          regulation of epithelial cell proliferation
          positive regulation of inflammatory response
          positive regulation of release of sequestered calcium ion into cytosol
          positive regulation of cytosolic calcium ion concentration involved in phospholipase C-activating G-protein coupled signaling pathway
          positive regulation of calcium ion transport
          maternal process involved in parturition
          head development
          positive regulation of ERK1 and ERK2 cascade
          cellular response to mechanical stimulus
          endothelin receptor signaling pathway
          positive regulation of neutrophil chemotaxis
          positive regulation of kidney development
          endothelin receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P158422 - -
          Agilent Whole Rat Genome Microarray 4x44K v2 A_64_P109657 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 544122 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 257197 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 686458 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 563737 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 249267 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 829517 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 208754 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543878 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 591070 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 144424 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 966210 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 96750 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 313117 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 497749 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 765665 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 691614 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 543383 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 57248 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 411981 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 438533 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 24694 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 413824 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 401997 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 858279 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 821103 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 934286 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608322 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 949999 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 378773 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 207393 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 188430 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 147709 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 960579 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 444629 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 961568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 12523 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 412659 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 608175 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 3167 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 881658 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 538401 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 198161 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 26872 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 47136 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 37320 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 119047 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 163766 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 160588 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 100975 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 132981 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158422 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P109657 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_64_P158422 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P158422 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_64_P109657 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 32261 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 39139 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 64386 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 95459 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 25520 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10807024 - -
          Affymetrix Rat Gene 1.0 ST Array 10807028 - -
          Affymetrix Rat Gene 1.0 ST Array 10807025 - -
          Affymetrix Rat Gene 1.0 ST Array 10807023 - -
          Affymetrix Rat Gene 1.0 ST Array 10807029 - -
          Affymetrix Rat Gene 1.0 ST Array 10807027 - -
          Affymetrix Rat Gene 1.0 ST Array 10807026 - -
          Affymetrix Rat Gene 1.0 ST Array 10807030 - -
          Affymetrix Rat Gene 1.1 ST Array 10807022 - -
          Rat Genome 230 2.0 Array 1369511_PM_at - -
          Rat Genome 230 2.0 Array 1369511_at - -
          Rat Genome 230 2.0 Array 1378342_PM_at - -
          Rat Genome 230 2.0 Array 1383641_at - -
          Rat Genome 230 2.0 Array 1393415_PM_at - -
          Rat Genome 230 2.0 Array 1393415_at - -
          Rat Genome 230 2.0 Array 1383641_PM_at - -
          Rat Genome 230 2.0 Array 1378342_at - -
          Affymetrix HT Rat Focus Array 1383641_PM_at - -
          Affymetrix HT Rat Focus Array 1369511_at - -
          Affymetrix HT Rat Focus Array 1383641_at - -
          Affymetrix HT Rat Focus Array 1378342_PM_at - -
          Affymetrix HT Rat Focus Array 1378342_at - -
          Affymetrix HT Rat Focus Array 1393415_at - -
          Affymetrix HT Rat Focus Array 1393415_PM_at - -
          Affymetrix HT Rat Focus Array 1369511_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1900003442.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003449.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003450.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003451.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003437.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003446.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003441.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003448.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003443.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003453.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003438.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003440.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003447.rn.1 TC1900000437.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR1900003452.rn.1 TC1900000437.rn.1 RTA TC
          Rat Expression Set 230 1369511_at - -
          Rat Expression Set 230 1369511_PM_at - -
          Rat Expression Set 230 1383641_at - -
          Rat Expression Set 230 1378342_at - -
          Rat Expression Set 230 1393415_PM_at - -
          Rat Expression Set 230 1383641_PM_at - -
          Rat Expression Set 230 1393415_at - -
          Rat Expression Set 230 1378342_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10807022 - -
          Rat Genome U34 Set X93210mRNA_at - -
          Rat Genome U34 Set M60786_at - -
          Rat Genome U34 Set rc_AA858906_at - -
          Rat Genome U34 Set rc_AI170069_at - -
          Rat Genome U34 Set X93210mRNA_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22193262 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Fair Trade Fair Trade
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Korea flag icon
          Korea

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담

           

          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat

          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910

           

          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783

           

          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)

          ISMS Logo

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline