Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561196_m1
          See other FGG GE Assays ›
          Gene Symbol:
          Fgg
          Gene Name:
          fibrinogen gamma chain
          Gene Aliases:
          -
          Chromosome Location:
          Chr.2: 181987080 - 181994523 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561196_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fgg
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012559.2 NP_036691.2 8 - 9 1160 89
          XM_006232525.1 XP_006232587.1 8 - 9 1314 89
          GenBank mRNA BC078893.1 - 8 - 9 1160 7122366 89
          FQ209468.1 - 4 - 5 633 89
          FQ210959.1 - 2 - 3 418 89
          FQ218666.1 - 6 - 7 962 89
          FQ219408.1 - 8 - 9 1185 89
          FQ219511.1 - 8 - 9 1182 89
          FQ219541.1 - 8 - 9 1181 89
          FQ219604.1 - 8 - 9 1184 89
          FQ229132.1 - 8 - 9 1187 89
          J00734.1 - 8 - 9 1145 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC0200001459.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fgg

          Panther Classification:

          Molecular Function -

          intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          inflammatory response
          cell-matrix adhesion
          signal transduction
          positive regulation of heterotypic cell-cell adhesion
          cellular protein complex assembly
          positive regulation of vasoconstriction
          positive regulation of exocytosis
          positive regulation of protein secretion
          protein polymerization
          response to calcium ion
          positive regulation of ERK1 and ERK2 cascade
          platelet aggregation
          positive regulation of peptide hormone secretion
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of endothelial cell apoptotic process
          molecular_function
          receptor binding
          structural molecule activity
          protein binding, bridging
          metal ion binding
          cell adhesion molecule binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11464 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 636450 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 340497 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 957539 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 432568 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 83766 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 511439 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11464 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11464 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_43_P11464 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_44_P558641 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 701431517 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 23706 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 79754 - -
          Agilent Rat cDNA Microarray (G4105A) 701431517 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10816068 - -
          Affymetrix Rat Gene 1.0 ST Array 10816071 - -
          Affymetrix Rat Gene 1.0 ST Array 10816070 - -
          Affymetrix Rat Gene 1.0 ST Array 10816074 - -
          Affymetrix Rat Gene 1.0 ST Array 10816069 - -
          Affymetrix Rat Gene 1.0 ST Array 10816078 - -
          Affymetrix Rat Gene 1.0 ST Array 10816072 - -
          Affymetrix Rat Gene 1.0 ST Array 10816076 - -
          Affymetrix Rat Gene 1.0 ST Array 10816073 - -
          Affymetrix Rat Gene 1.0 ST Array 10816075 - -
          Affymetrix Rat Gene 1.1 ST Array 10816067 - -
          Rat Genome 230 2.0 Array 1370086_PM_at - -
          Rat Genome 230 2.0 Array 1370086_at - -
          Affymetrix HT Rat Focus Array 1370086_PM_at - -
          Affymetrix HT Rat Focus Array 1370086_at - -
          Affymetrix Rat Clariom S Assay PSR0200008906.rn.1 TC0200001459.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200008905.rn.1 TC0200001459.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200008902.rn.1 TC0200001459.rn.1 RTA TC
          Affymetrix Rat Clariom S Assay PSR0200008903.rn.1 TC0200001459.rn.1 RTA TC
          Rat Expression Set 230 1370086_at - -
          Rat Expression Set 230 1370086_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10816067 - -
          Rat Genome U34 Set J00735_g_at - -
          Rat Genome U34 Set X05861exon#1-6_s_at - -
          Rat Genome U34 Set J00735_at - -
          Rat Genome U34 Set rc_AI169690_s_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 20754060 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1356909 - -
          RatRef-12 Expression BeadChip ILMN_48904 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline