Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Rn00561359_m1
          See other GRIN2C GE Assays ›
          Gene Symbol:
          Grin2c
          Gene Name:
          glutamate receptor, ionotropic, N-methyl D-aspartate 2C
          Gene Aliases:
          GluN2C, NR2C
          Chromosome Location:
          Chr.10: 103798290 - 103816287 on Build Rnor_6.0
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          Assay ID Rn00561359_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Grin2c
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_012575.3 NP_036707.3 12 - 13 2753 57
          XM_006247706.2 XP_006247768.1 10 - 11 2472 57
          XM_006247707.1 XP_006247769.1 13 - 14 2914 57
          XM_006247708.2 XP_006247770.1 10 - 11 2369 57
          XM_006247709.2 XP_006247771.1 10 - 11 2369 57
          GenBank mRNA D13212.1 - 12 - 13 2753 57
          M91563.1 - 10 - 11 2372 57
          U08259.1 - 13 - 14 2913 57

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as RTA transcript cluster(s) TC1000004104.rn.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Grin2c

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          startle response
          protein localization
          response to wounding
          directional locomotion
          ion transmembrane transport
          ionotropic glutamate receptor signaling pathway
          synaptic transmission, glutamatergic
          negative regulation of protein catabolic process
          regulation of membrane potential
          neuromuscular process controlling balance
          excitatory postsynaptic potential
          cation transmembrane transport
          ionotropic glutamate receptor activity
          NMDA glutamate receptor activity
          extracellular-glutamate-gated ion channel activity
          cation channel activity
          protein binding
          PDZ domain binding
          protein N-terminus binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Rat Genome Microarray 4x44K v2 A_43_P11471 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 517611 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 184048 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 392352 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 86774 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 629633 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 296873 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 734912 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 246454 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 227016 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 644893 - -
          Agilent SurePrint G3 Rat CGH Microarray 1x1M 393393 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 42774 - -
          Agilent SurePrint G3 Rat CGH Microarray 4x180K 5172 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11471 - -
          Agilent Whole Rat Genome Microarray 4x44K v3 A_43_P11471 - -
          Agilent SurePrint G3 Rat GE 8x60K Microarray A_43_P11471 - -
          SurePrint G3 Rat Gene Expression Microarray G4131A A_42_P738337 - -
          Matsumoto (Agilent Rat cDNA Microarray G4105A) 700327122 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 58988 - -
          Agilent Rat Genome CGH Microarray 105A (G4436A) 4217 - -
          Agilent Rat cDNA Microarray (G4105A) 700327122 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Rat Gene 1.0 ST Array 10748907 - -
          Affymetrix Rat Gene 1.0 ST Array 10748896 - -
          Affymetrix Rat Gene 1.0 ST Array 10748899 - -
          Affymetrix Rat Gene 1.0 ST Array 10748895 - -
          Affymetrix Rat Gene 1.0 ST Array 10748894 - -
          Affymetrix Rat Gene 1.0 ST Array 10748901 - -
          Affymetrix Rat Gene 1.0 ST Array 10748892 - -
          Affymetrix Rat Gene 1.0 ST Array 10748897 - -
          Affymetrix Rat Gene 1.0 ST Array 10748908 - -
          Affymetrix Rat Gene 1.0 ST Array 10748903 - -
          Affymetrix Rat Gene 1.0 ST Array 10748898 - -
          Affymetrix Rat Gene 1.0 ST Array 10748905 - -
          Affymetrix Rat Gene 1.0 ST Array 10748900 - -
          Affymetrix Rat Gene 1.0 ST Array 10748893 - -
          Affymetrix Rat Gene 1.0 ST Array 10748906 - -
          Affymetrix Rat Gene 1.0 ST Array 10748902 - -
          Affymetrix Rat Gene 1.0 ST Array 10748904 - -
          Affymetrix Rat Gene 1.1 ST Array 10748891 - -
          Rat Genome 230 2.0 Array 1368306_at - -
          Rat Genome 230 2.0 Array 1368306_PM_at - -
          Affymetrix HT Rat Focus Array 1368306_at - -
          Affymetrix HT Rat Focus Array 1368306_PM_at - -
          Affymetrix Rat Clariom S Assay PSR1000037186.rn.1 TC1000004104.rn.1 RTA TC
          Rat Expression Set 230 1368306_at - -
          Rat Expression Set 230 1368306_PM_at - -
          Affymetrix Rat Gene 1.0 ST Array 10748891 - -
          Rat Genome U34 Set U08259_r_at - -
          Rat Genome U34 Set D13212_s_at - -
          Rat Genome U34 Set D13212_s_at - -
          Rat Genome U34 Set U08259_r_at - -
          Array Name Probe IDs Targets Target Type
          ABI Rat Genome Survey Microarray 22365047 - -
          Array Name Probe IDs Targets Target Type
          Illumina ratRef-12 v1.0 expression beadchip ILMN_1358782 - -
          RatRef-12 Expression BeadChip ILMN_47797 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline