Search
Search
| Gene Symbol: Hmox1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012580.2 | NP_036712.1 | 2 - 3 | 263 | 132 | |
| GenBank mRNA | BC091164.1 | - | 2 - 3 | 263 | 7384448 | 132 |
| FQ228696.1 | - | 1 - 2 | 137 | 132 | ||
Set Membership: |
|